Search
View additional product information for Lambda DNA/EcoRI plus HindIII Marker - FAQs (SM0192, SM0191)
1 product FAQs found
Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).