Ladders can now be stored at room temperature for up to 24 months! During transition period you may receive packing indicating 6-month RT stability.
MassRuler DNA Ladders, ready-to-use
Thermo Scientific™

MassRuler DNA Ladders, ready-to-use

Thermo Scientific MassRuler DNA Ladders are designed for quantification and sizing of DNA fragments by agarose gel electrophoresis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize Range
SM03931500 to 10000 bp
SM038380 to 1031 bp
SM040380 to 10000 bp
SM1263100 to 1000 bp
SM1283100 to 1000 bp
Catalog number SM0393
Price (USD)
126.65
Online exclusive
169.00
Save 42.35
2 x 500 µL
Size Range:
1500 to 10000 bp
Request bulk or custom format
Price (USD)
126.65
Online exclusive
169.00
Save 42.35
2 x 500 µL

Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.

Highlights

  • Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months

Sustainability features

Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel CompatibilityAgarose Gels
Quantity2 x 500 μL
Ready to LoadYes
Shelf Life24 Months when stored at room temperature.
Gel TypeAgarose
Label or DyeBromophenol Blue
Product LineMassRuler
Product TypeHigh DNA Mass Ladder
Size Range1500 to 10000 bp
Unit Size2 x 500 µL

Frequently asked questions (FAQs)

Are the Thermo Scientific DNA ladders compatible with GelRed and GelGreen gel stains?

The GelRed or GelGreen nucleic acid dyes can be used with our DNA Ladders. Nucleic acid binding dyes can affect DNA migration during electrophoresis, therefore we highly recommend post-staining of gels. Please follow the gel staining protocol provided by the manufacturer of nucleic acid gel dyes.

Which Thermo Scientific DNA ladders are optimal for fast electrophoresis separation?

For fast sizing and quantification of double-stranded DNA, we recommend using FastRuler DNA Ladders. These ladders are easily resolved in a short separation distance after an 8-14 min run.

What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?

Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).

What are the end functional groups of Thermo Scientific DNA and RNA ladders?

Thermo Scientific DNA and RNA ladders have 5′-phosphate and 3′-hydroxyl groups.