NoLimits 10 bp DNA Fragment - FAQs
NoLimits 10 bp DNA Fragment - FAQs
View additional product information for NoLimits 10 bp DNA Fragment - FAQs (SM1391)
1 product FAQs found
Are all NoLimits DNA Fragments (range: 10 - 20,000 bp; Cat. No. SM1391) double-stranded?
Yes, all NoLimits DNA Fragments (Cat. No. SM1391) are, from 10 - 20,000 bp, double-stranded.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Gel Electrophoresis and Blotting Support Center.