Thermo Scientific™
Smart-Vue™ 915MHz Wireless Monitoring Solution Kits (North America)
Catalog number: SMARTKIT3A
Thermo Scientific™
Smart-Vue™ 915MHz Wireless Monitoring Solution Kits (North America)
Catalog number: SMARTKIT3A
Protect sample quality and facilitate regulatory compliance with Thermo Scientific™ Smart-Vue™ 915MHz Wireless Monitoring Solution Kits (North America). These solutions provide continuous monitoring and reporting of your laboratory equipment with immediate remote alarm notification and continuous data collection.
Catalog Number
SMARTKIT3A
Unit Size
Each
For Use With
Incubators (temperature only)
Includes
Five calibrated wireless modules with PT100 sensors
Price (USD)
Full specifications
For Use WithIncubators (temperature only)
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
Data Memory3000 Readings
Display TypeLCD
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
DescriptionSmart-Vue Kit for Incubators; w/o Client server software and USB receiver
Temperature Range (Metric)-200°C to +350°C
IncludesFive calibrated wireless modules with PT100 sensors
CalibrationCalibrated with Wireless Module
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Product TypeWireless Monitoring Solution Kit
TypeWireless Temperature Module with Sensor
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With
ULT Freezers
Includes
Five calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver
Price (USD)
Full specifications
For Use WithULT Freezers
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
Data Memory3000 Readings
Display TypeLCD
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
DescriptionSmart-Vue kit of 5 calibrated wireless modules for ULT freezers
Temperature Range (Metric)-200°C to +350°C
IncludesFive calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver
CalibrationCalibrated with Wireless Module
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Product TypeWireless Monitoring Solution Kit
TypeWireless Temperature Module with Sensor
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With
ULT Freezers
Includes
Five calibrated wireless modules with PT100 sensors
Price (USD)
Full specifications
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
DescriptionSmart-Vue Kit for ULT Freezers; w/o Client server software and USB receiver
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
CalibrationCalibrated with Wireless Module
Data Memory3000 Readings
Display TypeLCD
Product TypeWireless Monitoring Solution Kit
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
Temperature Range (Metric)-200°C to +350°C
For Use WithULT Freezers
IncludesFive calibrated wireless modules with PT100 sensors
TypeWireless Temperature Module with Sensor
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With
Refrigerators
Includes
Five calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver
Price (USD)
Full specifications
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
DescriptionSmart-Vue kit of 5 calibrated wireless modules for refrigerators
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
CalibrationCalibrated with Wireless Module
Data Memory3000 Readings
Display TypeLCD
Product TypeWireless Monitoring Solution Kit
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
Temperature Range (Metric)-200°C to +350°C
For Use WithRefrigerators
IncludesFive calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver
TypeWireless Temperature Module with Sensor
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With
Refrigerators
Includes
Five calibrated wireless modules with PT100 sensors
Price (USD)
Full specifications
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
DescriptionSmart-Vue Kit for Refrigerators; w/o Client server software and USB receiver
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
CalibrationCalibrated with Wireless Module
Data Memory3000 Readings
Display TypeLCD
Product TypeWireless Monitoring Solution Kit
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
Temperature Range (Metric)-200°C to +350°C
For Use WithRefrigerators
IncludesFive calibrated wireless modules with PT100 sensors
TypeWireless Temperature Module with Sensor
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With
Incubators (temperature only)
Includes
Five calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver
Price (USD)
Full specifications
For Use WithIncubators (temperature only)
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
Data Memory3000 Readings
Display TypeLCD
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
DescriptionSmart-Vue kit of 5 calibrated wireless modules for incubator
Temperature Range (Metric)-200°C to +350°C
IncludesFive calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver
CalibrationCalibrated
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Product TypeWireless Monitoring Solution Kit
TypeWireless Temperature Module with Sensor
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeFor Use WithIncludesPrice (USD)
SMARTKIT3AFull specifications
EachIncubators (temperature only)Five calibrated wireless modules with PT100 sensorsRequest A Quote
For Use WithIncubators (temperature only)
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
Data Memory3000 Readings
Display TypeLCD
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
DescriptionSmart-Vue Kit for Incubators; w/o Client server software and USB receiver
Temperature Range (Metric)-200°C to +350°C
IncludesFive calibrated wireless modules with PT100 sensors
CalibrationCalibrated with Wireless Module
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Product TypeWireless Monitoring Solution Kit
TypeWireless Temperature Module with Sensor
Unit SizeEach
SMARTKIT1Full specifications
EachULT FreezersFive calibrated wireless modules with PT100 sensors, client server software and 1 USB receiverRequest A Quote
For Use WithULT Freezers
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
Data Memory3000 Readings
Display TypeLCD
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
DescriptionSmart-Vue kit of 5 calibrated wireless modules for ULT freezers
Temperature Range (Metric)-200°C to +350°C
IncludesFive calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver
CalibrationCalibrated with Wireless Module
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Product TypeWireless Monitoring Solution Kit
TypeWireless Temperature Module with Sensor
Unit SizeEach
SMARTKIT1AFull specifications
EachULT FreezersFive calibrated wireless modules with PT100 sensorsRequest A Quote
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
DescriptionSmart-Vue Kit for ULT Freezers; w/o Client server software and USB receiver
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
CalibrationCalibrated with Wireless Module
Data Memory3000 Readings
Display TypeLCD
Product TypeWireless Monitoring Solution Kit
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
Temperature Range (Metric)-200°C to +350°C
For Use WithULT Freezers
IncludesFive calibrated wireless modules with PT100 sensors
TypeWireless Temperature Module with Sensor
Unit SizeEach
SMARTKIT2Full specifications
EachRefrigeratorsFive calibrated wireless modules with PT100 sensors, client server software and 1 USB receiverRequest A Quote
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
DescriptionSmart-Vue kit of 5 calibrated wireless modules for refrigerators
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
CalibrationCalibrated with Wireless Module
Data Memory3000 Readings
Display TypeLCD
Product TypeWireless Monitoring Solution Kit
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
Temperature Range (Metric)-200°C to +350°C
For Use WithRefrigerators
IncludesFive calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver
TypeWireless Temperature Module with Sensor
Unit SizeEach
SMARTKIT2AFull specifications
EachRefrigeratorsFive calibrated wireless modules with PT100 sensorsRequest A Quote
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
DescriptionSmart-Vue Kit for Refrigerators; w/o Client server software and USB receiver
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
CalibrationCalibrated with Wireless Module
Data Memory3000 Readings
Display TypeLCD
Product TypeWireless Monitoring Solution Kit
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
Temperature Range (Metric)-200°C to +350°C
For Use WithRefrigerators
IncludesFive calibrated wireless modules with PT100 sensors
TypeWireless Temperature Module with Sensor
Unit SizeEach
SMARTKIT3Full specifications
EachIncubators (temperature only)Five calibrated wireless modules with PT100 sensors, client server software and 1 USB receiverRequest A Quote
For Use WithIncubators (temperature only)
Alarm TypesTelephone, Email, SMS Text, Fax, Audible, Visual, Printed Report
Battery Type3.6 V Lithium
Data Memory3000 Readings
Display TypeLCD
Sensor TypesDifferential Pressure Sensor, CO2 Sensor
Software TypeSmart-Vue Client Software, Smart-Vue Server Software
DescriptionSmart-Vue kit of 5 calibrated wireless modules for incubator
Temperature Range (Metric)-200°C to +350°C
IncludesFive calibrated wireless modules with PT100 sensors, client server software and 1 USB receiver
CalibrationCalibrated
Certifications/ComplianceSmart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Product TypeWireless Monitoring Solution Kit
TypeWireless Temperature Module with Sensor
Unit SizeEach
Showing 6 of 6
Sample Protection and Security
  • Permanently records all readings, alarms, and user acknowledgments
  • Protects work 24 hours a day, seven days a week with immediate remote alarm notification and continuous data collection
  • Audit trail traceability assists with conformance to 21 CFR part 11 and other regulatory requirements
  • Monitors temperatures ranging from -200° to +350°C, CO2 concentration, relative humidity (RH), differential pressure (clean rooms), 4-20 milliamps and dry contact state
Simplicity
  • Easy to install, use and maintain
  • No complicated wiring, allowing easy movement of laboratory equipment
  • Accessible anytime, anywhere via the internet with Smart-Vue client software
  • Quickly export reports to Microsoft™ Excel™, Word™ or Adobe™ PDF formats with just a click for auditing purposes
  • View multiple sensors
  • Alarm options: telephone, email, SMS text, fax, audible, visual and printed reports
Scalable
  • Single software solution, that is scalable and will grow with you, saving time and money
  • Easy to add new laboratory equipment to an existing Smart-Vue system
Complementary Services
  • Installation and validation services
  • Annual maintenance agreements
  • Calibration, inspection, battery replacement and software updates
  • Re-qualification (validation) as needed for cGxP compliance
Ordering Information:
Similar packaged solutions are available for cryopreservation systems, ovens, CO2 incubators, environmental chambers, water baths and other equipment. Smart-Vue modules and sensors are available for temperature, RH, CO2 concentration, differential pressure, 4-20mA output, ambient conditions, and alarm/relay dry contacts. Solutions vary by RF regions worldwide and are compatible with multiple brands and types of laboratory equipment. Please ask your Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements.
Compatible with:
Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
Recommended for:
For use in highly controlled environments, including pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories.
Note:
Some incubators have a decontamination feature; please note that the SMARTKIT3 is not designed to monitor temperatures over 120°C.
The purchase of Smart-Vue requires software installation, contact your sales representative for details.
Figures