Thermo Scientific M13/pUC sequencing primers anneal to the region in the 5'-terminus of the lacZ gene. All primers are supplied as 10 µM aqueous solutions.
Applications
• Sequencing of DNA fragments inserted into the MCS within the lacZ gene of various cloning vectors, such as pUC19, pTZ19R, pTZ57R, M13mp18, and pBluescript II • Colony screening by PCR
For Research Use Only. Not for use in diagnostic procedures.