Thermo Scientific™

M13/pUC Reverse Sequencing Primer (-46), 24-mer

Catalog number: SO115
Thermo Scientific™

M13/pUC Reverse Sequencing Primer (-46), 24-mer

Catalog number: SO115
Catalog Number
SO115
Unit Size
10 µM
Price (USD)
Price: 80.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SO11510 µM
Price: 80.00
Your Price:
-

Recurring order eligible

Save time by having your items shipped automatically.

Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific M13/pUC sequencing primers anneal to the region in the 5'-terminus of the lacZ gene. All primers are supplied as 10 µM aqueous solutions.

Applications

• Sequencing of DNA fragments inserted into the MCS within the lacZ gene of various cloning vectors, such as pUC19, pTZ19R, pTZ57R, M13mp18, and pBluescript II
• Colony screening by PCR
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Sequencing Primer
Primer
M13
Vector
pUC19, pTZ19R, pTZ57R, M13mp18, pBluescript II
Form
Liquid