Oligo(dT)<sub>18</sub> Primer
Thermo Scientific™

Oligo(dT)18 Primer

Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied asRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
SO132120 μL
SO13160 μL
Catalog number SO132
Price (TWD)
5,580.00
Online offer
Ends: 30-Jun-2026
7,440.00
Save 1,860.00 (25%)
120 µL
Quantity:
120 μL
Price (TWD)
5,580.00
Online offer
Ends: 30-Jun-2026
7,440.00
Save 1,860.00 (25%)
120 µL
Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready-to-use, 20X concentrated aqueous solution.

Applications
• cDNA Synthesis
• RT-qPCR
• RT-PCR

Related products
Anchored Oligo dT
Random Hexamer Primer
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)cDNA Synthesis
FormLiquid
Primer Length18-mer
Product TypePrimer
Quantity120 μL
Shipping ConditionDry Ice
Concentration0.5 μg/μL
PrimerOligo dT
Unit Size120 µL
Contents & Storage
• Oligo(dT)18 Primer (120 μL at 0.5 μg/μL)

Store at –20°C.