Thermo Scientific™

Dionex™ CarboPac™ PA1 IC Columns

Catalog number: SP2866
Thermo Scientific™

Dionex™ CarboPac™ PA1 IC Columns

Catalog number: SP2866
For isocratic determinations of monosaccharides, disaccharides, and linear polysaccharides in a variety of matrices, use the rugged Thermo Scientific™ Dionex™ CarboPac™ PA1 Analytical & Guard Columns. This specialized anion-exchange column, combined with pulsed amperometric detection (PAD), delivers high-resolution separations. The unique Thermo Scientific™ Dionex™ MicroBead™ pellicular structure of the Dionex CarboPac PA1 resin gives it stability from pH 0–14 at all concentrations of buffer salts. The pellicular resin structure provides excellent mass transfer resulting in fast gradient re-equilibration.
 
Catalog Number
SP2866
Unit Size
Each 1
Type
Preparative Column
Diameter (Metric)
22 mm
Length (Metric)
250 mm
Price (USD)
7,514.84
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity3025 µeq
For Use With (Application)Separation of Mono, Di, and Specific Oligosaccharides
For Use With (Equipment)Separation of Mono, Di and Specific Oligosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents, Acetate or Hydroxide Eluents Only
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypePreparative Column
pH0 to 14
Length (Metric)250 mm
Diameter (Metric)22 mm
Temperature (Metric)4°C to 60°C
Column ConstructionPEEK
Resin Composition10 µm-Diameter Substrate (Polystyrene 2% Cross-Linked with Divinylbenzene) Agglomerated with 500 nm MicroBead Quaternary Ammonium Functionalized Latex (5% Cross-Linked).
Unit SizeEach 1
Catalog Number
Unit Size
Each 1
Type
Microbore Analytical Column
Diameter (Metric)
2 mm
Length (Metric)
250 mm
Price (USD)
1,415.67
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity25 µeq
Flow Rate1.0 mL/min.
For Use With (Application)Carbohydrate Analysis
For Use With (Equipment)PAD Systems
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents, Acetate or Hydroxide Eluents Only
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeMicrobore Analytical Column
pH0 to 14
Length (Metric)250 mm
Diameter (Metric)2 mm
Temperature (Metric)4°C to 60°C
Column ConstructionPEEK
Resin Composition10 µm-Diameter Substrate (Polystyrene 2% Cross-Linked with Divinylbenzene) Agglomerated with 500 nm MicroBead Quaternary Ammonium Functionalized Latex (5% Cross-Linked).
Unit SizeEach 1
Catalog Number
Unit Size
Each 1
Type
Microbore Guard Column
Diameter (Metric)
2 mm
Length (Metric)
50 mm
Price (USD)
469.58
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity5 µeq
For Use With (Application)Separation of Neutral and Acidic Monosaccharides
For Use With (Equipment)Separation of Neutral, Acidic Monosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeMicrobore Guard Column
Length (Metric)50 mm
Diameter (Metric)2 mm
Unit SizeEach 1
Catalog Number
Unit Size
Each 1
Type
Standard Bore Guard Column
Diameter (Metric)
4 mm
Length (Metric)
50 mm
Price (USD)
469.58
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity20 µeq
For Use With (Application)Separation of Neutral and Acidic Monosaccharides
For Use With (Equipment)Separation of Neutral, Acidic Monosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeStandard Bore Guard Column
pH0 to 14
Length (Metric)50 mm
Diameter (Metric)4 mm
Unit SizeEach 1
Catalog Number
Unit Size
Each 1
Type
Semi-Preparative Column
Diameter (Metric)
9 mm
Length (Metric)
250 mm
Price (USD)
2,818.07
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity500 µeq
For Use With (Application)Separation of Mono, Di, and Specific Oligosaccharides
For Use With (Equipment)Separation of Mono, Di and Specific Oligosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents, Acetate or Hydroxide Eluents Only
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeSemi-Preparative Column
pH0 to 14
Length (Metric)250 mm
Diameter (Metric)9 mm
Temperature (Metric)4°C to 60°C
Column ConstructionPEEK
Resin Composition10 µm-Diameter Substrate (Polystyrene 2% Cross-Linked with Divinylbenzene) Agglomerated with 500 nm MicroBead Quaternary Ammonium Functionalized Latex (5% Cross-Linked).
Unit SizeEach 1
Catalog Number
Unit Size
Each 1
Type
Semi-Preparative Guard Column
Diameter (Metric)
9 mm
Length (Metric)
50 mm
Price (USD)
1,216.42
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity100 µeq
For Use With (Application)Separation of Mono, Di, and Specific Oligosaccharides
For Use With (Equipment)Separation of Mono, Di and Specific Oligosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeSemi-Preparative Guard Column
pH0 to 14
Length (Metric)50 mm
Diameter (Metric)9 mm
Unit SizeEach 1
Catalog Number
Unit Size
Each 1
Type
Standard Bore Analytical Column
Diameter (Metric)
4 mm
Length (Metric)
250 mm
Price (USD)
1,415.67
Your Price:
Availability
***
Full specifications
Quantity
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity100 µeq
Flow Rate1.0 mL/min.
For Use With (Application)Carbohydrate Analysis
For Use With (Equipment)ICS-2000, ICS-3000 IC Systems
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents, Acetate or Hydroxide Eluents Only
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeStandard Bore Analytical Column
pH0 to 14
Length (Metric)250 mm
Diameter (Metric)4 mm
Temperature (Metric)4°C to 60°C
Column ConstructionPEEK
Resin Composition10 µm-Diameter Substrate (Polystyrene 2% Cross-Linked with Divinylbenzene) Agglomerated with 500 nm MicroBead Quaternary Ammonium Functionalized Latex (5% Cross-Linked).
Unit SizeEach 1
Showing 7 of 7
Catalog NumberSpecificationsUnit SizeTypeDiameter (Metric)Length (Metric)Price (USD)AvailabilityQuantity
SP2866Full specifications
Each 1Preparative Column22 mm250 mm
7,514.84
Your Price:
***
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity3025 µeq
For Use With (Application)Separation of Mono, Di, and Specific Oligosaccharides
For Use With (Equipment)Separation of Mono, Di and Specific Oligosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents, Acetate or Hydroxide Eluents Only
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypePreparative Column
pH0 to 14
Length (Metric)250 mm
Diameter (Metric)22 mm
Temperature (Metric)4°C to 60°C
Column ConstructionPEEK
Resin Composition10 µm-Diameter Substrate (Polystyrene 2% Cross-Linked with Divinylbenzene) Agglomerated with 500 nm MicroBead Quaternary Ammonium Functionalized Latex (5% Cross-Linked).
Unit SizeEach 1
057178Full specifications
Each 1Microbore Analytical Column2 mm250 mm
1,415.67
Your Price:
***
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity25 µeq
Flow Rate1.0 mL/min.
For Use With (Application)Carbohydrate Analysis
For Use With (Equipment)PAD Systems
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents, Acetate or Hydroxide Eluents Only
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeMicrobore Analytical Column
pH0 to 14
Length (Metric)250 mm
Diameter (Metric)2 mm
Temperature (Metric)4°C to 60°C
Column ConstructionPEEK
Resin Composition10 µm-Diameter Substrate (Polystyrene 2% Cross-Linked with Divinylbenzene) Agglomerated with 500 nm MicroBead Quaternary Ammonium Functionalized Latex (5% Cross-Linked).
Unit SizeEach 1
057179Full specifications
Each 1Microbore Guard Column2 mm50 mm
469.58
Your Price:
***
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity5 µeq
For Use With (Application)Separation of Neutral and Acidic Monosaccharides
For Use With (Equipment)Separation of Neutral, Acidic Monosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeMicrobore Guard Column
Length (Metric)50 mm
Diameter (Metric)2 mm
Unit SizeEach 1
043096Full specifications
Each 1Standard Bore Guard Column4 mm50 mm
469.58
Your Price:
***
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity20 µeq
For Use With (Application)Separation of Neutral and Acidic Monosaccharides
For Use With (Equipment)Separation of Neutral, Acidic Monosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeStandard Bore Guard Column
pH0 to 14
Length (Metric)50 mm
Diameter (Metric)4 mm
Unit SizeEach 1
039686Full specifications
Each 1Semi-Preparative Column9 mm250 mm
2,818.07
Your Price:
***
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity500 µeq
For Use With (Application)Separation of Mono, Di, and Specific Oligosaccharides
For Use With (Equipment)Separation of Mono, Di and Specific Oligosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents, Acetate or Hydroxide Eluents Only
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeSemi-Preparative Column
pH0 to 14
Length (Metric)250 mm
Diameter (Metric)9 mm
Temperature (Metric)4°C to 60°C
Column ConstructionPEEK
Resin Composition10 µm-Diameter Substrate (Polystyrene 2% Cross-Linked with Divinylbenzene) Agglomerated with 500 nm MicroBead Quaternary Ammonium Functionalized Latex (5% Cross-Linked).
Unit SizeEach 1
063501Full specifications
Each 1Semi-Preparative Guard Column9 mm50 mm
1,216.42
Your Price:
***
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity100 µeq
For Use With (Application)Separation of Mono, Di, and Specific Oligosaccharides
For Use With (Equipment)Separation of Mono, Di and Specific Oligosaccharides
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeSemi-Preparative Guard Column
pH0 to 14
Length (Metric)50 mm
Diameter (Metric)9 mm
Unit SizeEach 1
035391Full specifications
Each 1Standard Bore Analytical Column4 mm250 mm
1,415.67
Your Price:
***
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity100 µeq
Flow Rate1.0 mL/min.
For Use With (Application)Carbohydrate Analysis
For Use With (Equipment)ICS-2000, ICS-3000 IC Systems
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents, Acetate or Hydroxide Eluents Only
Particle Size10 µm
Stationary PhaseCarboPac PA1
TypeStandard Bore Analytical Column
pH0 to 14
Length (Metric)250 mm
Diameter (Metric)4 mm
Temperature (Metric)4°C to 60°C
Column ConstructionPEEK
Resin Composition10 µm-Diameter Substrate (Polystyrene 2% Cross-Linked with Divinylbenzene) Agglomerated with 500 nm MicroBead Quaternary Ammonium Functionalized Latex (5% Cross-Linked).
Unit SizeEach 1
Showing 7 of 7
Durable Column for Carbohydrate Separations
  • Approved for use in a variety of official methods for the analysis of foods
  • Well suited for the analysis of foods for nutritional labeling
  • Ideal for the separation of neutral and acidic monosaccharides
  • Lower sensitivity than the Dionex CarboPac PA10 or PA20 Columns
  • Available in microbore, standard bore, and semipreparative formats

About the Dionex CarboPac Family of Columns

The Dionex CarboPac family offers a selection of high-performance liquid chromatography (HPLC) columns, each optimized for a different class of compounds. Combined with PAD, which provides a means to detect carbohydrates without derivatization, these columns support simple, reliable techniques to separate sugar alcohols, mono- and disaccharides, and poly- and oligosaccharides.

Dionex CarboPac columns provide high-resolution separations of:

  • Glycoprotein oligosaccharides and complex carbohydrates from dietary fiber, including fructans, maltodextrins, and amylopectins
  • Monosaccharides, such as those obtained from glycoprotein hydrolysates or plant hydrolysates
  • Mono- and disaccharides
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Frequently asked questions (FAQs)

Citations & References