Spinnaker Microplate Robot - FAQs
Spinnaker Microplate Robot - FAQs
View additional product information for Spinnaker Microplate Robot - FAQs (SPK0001)
1 product FAQs found
Are the Multidrop Combi reagent dispensers compatible with Momentum Workflow Scheduling Software?
Yes, the Multidrop Combi reagent dispensers are compatible with the Momentum Software and can be integrated into automated workflow platforms. If you are planning to set up an automated workflow using the Momentum Software, you will also need a microplate mover such as the Orbitor RS2 Microplate Mover, Spinnaker Microplate Robot, or Spinnaker 3 Microplate Mover.