Are ultra thin wall (UTW) vessels prone to break more often because of their thinner walls?
When used as instructed, these vessels are remarkably robust and can handle typical laboratory conditions, without fracturing or resulting in loss of volume. It is recommended that, when applying caps, a proper tray be used to protect the bottom of the tubes from excessive force that is typically used.
Find additional tips, troubleshooting help, and resources within our
Real-Time PCR and Digital PCR Instruments Support Center.
How do I know whether I need to purchase the frames for my Piko PCR Plates?
The frames for the Piko PCR Plates allow for easier handling and storage, particularly if you wish to use these plates in conjunction with laboratory equipment designed for microplates (such as centrifuges, plate readers or liquid handlers). Also, the frames allow the ability to stack the plates in a stable and compact fashion for archiving or storage.
What is a Piko PCR Plate?
A Piko PCR Plate is a plate for PCR that is equivalent to one-fourth of a standard microplate and is equal to a microscope slide in footprint. These plates conserve all of the standard well-spacing and well-geometry of conventional microplates, e.g., a 96-well Piko PCR Plate has the same 4.5 mm pitch as most conventional 384-well microplates. Up to four Piko PCR Plates can be reversibly snapped into a frame to give the equivalent footprint of a conventional 96-well or 384-well microplate. Piko PCR Plates provide many advantages over traditional microplates: they are available at a lower cost, they accept lower reaction volumes, and require less storage space. In addition, many of the same types of advantages were incorporated in the instrumentation designed to use these plates: the thermal cycler has a smaller footprint, is less expensive, has higher thermal performance specifications, and consumes less power.
Can I use full-height as well as low-profile PCR vessels in the Piko 24?
The 24-well Piko will only accept low-profile 0.2 mL tubes or plates with flat-caps or sealers. The taller design of vessels simply will not fit in the instrument. The taller design of full-height tubes was originally designed for centrifugation, not for PCR. Low-profile tubes and plates are optimal for PCR because the portion protruding above the cycler block has been shortened, minimizing the potential for condensation and reducing mass to allow faster thermal transfer.