Oxoid™ Blaser-Wang Campylobacter Selective Supplement
Thermo Scientific™
Oxoid™ Blaser-Wang Campylobacter Selective Supplement
Oxoid Campylobacter Selective Supplement, Blaser-Wang isolatee Campylobacter and inhibit Candida albicans.
Have Questions?
Catalog NumberQuantity
SR0098E10/Pk.
Catalog number SR0098E
Price (KRW)
-
Quantity:
10/Pk.
Oxoid Campylobacter Selective Supplement, Blaser-Wang is used to prepare of Blaser-Wang Selective Medium for the isolation of Campylobacter while inhibiting growth of Candida albicans.
  • Add to Columbia Blood Agar Base, Part No. CM0271B
  • Each vial supplements 500mL of medium
Specifications
DescriptionCampylobacter Selective Supplement, Blaser-Wang
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesCampylobacter
Unit SizeEach