Thermo Scientific™
Oxoid™ VCN Selective Supplement
Related applications:
Industrial Microbiology
Oxoid VCN Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
Back to top
  • Add to GC Agar Base, Part No. CM0367B
  • Each vial supplements 500mL of medium