Oxoid™ CCDA Selective Supplement
Imagem ilustrativa. O produto pode variar
Thermo Scientific™

Oxoid™ CCDA Selective Supplement

Oxoid Brain Heart Infusion Broth is highly nutritious medium for the growth of fastidious organisms, recommended for blood culture work; also see Heart Infusion Broth.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantityYield
SR0155H10 VialsFor 2L medium
SR0155E10/Pk.Each vial for 500mL medium
Número do catálogo SR0155H
Preço (BRL)
-
Quantity:
10 Vials
Yield:
For 2L medium

Oxoid CCDA Selective Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Arcobacter.

  • For the isolation of Arcobacter spp., add to Arcobacter Broth Base, Part No. CM0965B
  • For the isolation of Campylobacter spp., especially C. upsaliensis, add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B
Especificações
DescriptionCCDA Selective Supplement
Quantity10 Vials
YieldFor 2L medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach