Oxoid™ MRSV Selective Supplement
Thermo Scientific™
Oxoid™ MRSV Selective Supplement
Oxoid MRSV Selective Supplement is used for the detection of Salmonella spp. from food and environmental samples.
製品番号(カタログ番号)数量
SR0161E10/Pk.
製品番号(カタログ番号) SR0161E
価格(JPY)
-
数量:
10/Pk.
Oxoid MRSV Selective Supplement is used for the detection of Salmonella spp. from food and environmental samples.
  • Add to MSRV Medium Base, Part No. CM0910B and Lysine Iron Agar, Modified (MLIA), Part No. CM0886B
  • Each vial supplements 500mL of medium
仕様
概要MSRV Selective Supplement
数量10/Pk.
収量Each vial for 500mL medium
形状Freeze-dried
製品タイプSelective Supplement
Unit SizeEach