Oxoid™ C-T Supplement
Oxoid™ C-T Supplement
Actual product may vary
Thermo Scientific™

Oxoid™ C-T Supplement

Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.
Have Questions?
카탈로그 번호수율
SR0172EEach vial for 500mL medium
카탈로그 번호 SR0172E
제품 가격(KRW)
-
수율:
Each vial for 500mL medium

Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.

Add to Sorbitol MacConkey Agar, Part No. CM0813B or to Sorbitol MacConkey Agar with BCIG, Part No. CM0981B.

Ordering Information:

Each vial supplements 2L of medium.

사양
설명C-T Supplement
수량10/Pk.
수율Each vial for 500mL medium
형태Freeze-dried
제품 유형Microbiology Media
Unit SizeEach