Oxoid™ C-T Supplement
Actual product may vary
Thermo Scientific™

Oxoid™ C-T Supplement

Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberYield
SR0172HEach vial for 2L medium
SR0172EEach vial for 500mL medium
Catalog number SR0172H
Price (BRL)
-
Yield:
Each vial for 2L medium

Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.

Add to Sorbitol MacConkey Agar, Part No. CM0813B or to Sorbitol MacConkey Agar with BCIG, Part No. CM0981B.

Ordering Information:

Each vial supplements 2L of medium.

Specifications
DescriptionC-T Supplement
Quantity10/Pk.
YieldEach vial for 2L medium
FormFreeze-dried
Product TypeMicrobiology Media
Unit SizeEach