Thermo Scientific™
Oxoid™ Legionella BCYE Supplement
Related applications:
Industrial Microbiology
Oxoid Legionella BCYE Supplement w/o L-cysteine is used for the isolation of Legionella spp.
Back to top
  • Add to Legionella CYE Agar Base, Part No. CM0655B
  • Each vial supplements 100mL of medium