Oxoid™ ORSAB Selective Supplement
Thermo Scientific™
Oxoid™ ORSAB Selective Supplement
Oxoid ORSAB Selective Supplement (Oxacillin Resistance Screening Agar Base Selective Supplement) is added to Oxacillin Resistance Screening Agar Base, Part No. CM1008B.
Have Questions?
Catalog NumberQuantity
SR0195E10/Pk.
Catalog number SR0195E
Price (MXN)
-
Quantity:
10/Pk.
Oxoid ORSAB Selective Supplement (Oxacillin Resistance Screening Agar Base Selective Supplement) is added to Oxacillin Resistance Screening Agar Base, Part No. CM1008B.
  • Add to Oxacillin Resistance Screening Agar Base, Part No. CM1008B
  • Each vial supplements 500mL of medium
Specifications
DescriptionORSAB Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach