Product Image
Oxoid UK

TTC Solution 1% 10x5mL

For addition to K-F Streptococcus Agar (CM0701) for the isolation and enumeration of Enterococci.
Have Questions?
Catalog NumberQuantity
SR0229K10 x 5mL
Catalog number SR0229K
Price (MXN)
-
Quantity:
10 x 5mL
Oxoid™ TTC Solution is used for the enumeration and identification of coliform bacteria.
For Laboratory Use
For Laboratory Use
Specifications
Country of OriginUK
For Use With (Equipment)Supplement for K-F Streptococcus Agar (CM0701)
Media TypeSupplement
Packaging Type10 x 5 mL
Quantity10 x 5mL
Shelf Life18 months
YieldEach vial for 500mL medium
FormLiquid
Product TypeSolution
SpeciesEnterococci
Unit SizeEach
Contents & Storage
2°C to 8°C