Oxoid™ ONE Broth-Listeria Supplement
Thermo Scientific™

Oxoid™ ONE Broth-Listeria Supplement

The Oxoid ONE Broth-Listeria is for the selective enrichment broth for Listeria species from food samples in 24 hours.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수율
SR0234EEach vial for 500mL medium
SR0234BEach vial for 2.25L medium
SR0234HEach vial for 2 L medium
카탈로그 번호 SR0234E
제품 가격(KRW)
-
수율:
Each vial for 500mL medium

The Oxoid ONE Broth-Listeria is for the selective enrichment broth for Listeria species from food samples in 24 hours.

Add to ONE Broth-Listeria, Part No. CM1066.

ONE Broth - Listeria: Typical Formulation

  • Peptone: 26g/L; Carbohydrate mix: 6g/L; Salt mix: 10g/L with a final pH of 7.4 ±0.2 at 25°C

Appearance:

  • Dehydrated medium: straw coloured, free-flowing powder
  • Selective supplement: orange/yellow, freeze-dried pellet
  • Prepared medium: dark straw coloured, clear liquid

Note:

Formulation Directions

  • Suspend 22g of ONE Broth-Listeria Base in 500mL (44g/L) of distilled water
  • Mix well and sterilize by autoclaving at 121°C for 15 minutes
  • Cool the medium to below 50°C and aseptically add the contents of one vial of ONE Broth-Listeria Selective Supplement (Mfr. No. SR0234) reconstituted as directed

Testing Protocol

  • Add 25g of food sample to 225mL of ONE Broth-Listeria and stomach for a minimum of 30 seconds to mix the sample
  • Incubate the broth without agitation at 30°C for 24±2 hours
  • Gently agitate the bag then, using a microbiological loop, remove 10μL and inoculate onto a suitable agar medium e.g. Oxoid Brilliance Listeria Agar (ISO) (Mfr. No. CM1212, SR257 and SR0258) and incubate appropriately (37°C for 24-48 hours)

  • Confirm presumptive colonies on the agar plates as Listeria monocytogenes or Listeria spp. by appropriate methods e.g. Gram-stain, catalase, oxidase, Oxoid Listeria Latex Test Kit (Mfr. No. DR1126)
  • Oxoid O.B.I.S. mono (Mfr. No. ID0600), Microbact Listeria 12L (Mfr. No. MB1128)

 


For Laboratory Use Only

Formulation Directions

  • Suspend 22g of ONE Broth-Listeria Base in 500mL (44g/L) of distilled water
  • Mix well and sterilize by autoclaving at 121°C for 15 minutes
  • Cool the medium to below 50°C and aseptically add the contents of one vial of ONE Broth-Listeria Selective Supplement (Mfr. No. SR0234) reconstituted as directed

Testing Protocol

  • Add 25g of food sample to 225mL of ONE Broth-Listeria and stomach for a minimum of 30 seconds to mix the sample
  • Incubate the broth without agitation at 30°C for 24±2 hours
  • Gently agitate the bag then, using a microbiological loop, remove 10μL and inoculate onto a suitable agar medium e.g. Brilliance Listeria Agar (Mfr. No. CM1080, SR0222, and SR0228) and incubate appropriately (37°C for 24 to 48 hours)
  • Confirm presumptive colonies on the agar plates as Listeria monocytogenes or Listeria spp. by appropriate methods e.g. Gram-stain, catalase, oxidase, Oxoid Listeria Latex Test Kit DR1126, Oxoid O.B.I.S. mono ID0600, Microbact Listeria 12L MB1128

General References:

  1. ISO 11290-1 (Microbiology of food and animal feeding stuffs — Horizontal method for the enumeration of Listeria monocytogenes — part 1).
  2. Oxoid Poster - Folio No. 1033, July 2004.
  3. Data on file at Oxoid
사양
설명ONE Broth-Listeria Supplement
수량10/Pk.
수율Each vial for 500mL medium
형태Freeze-dried
제품 유형Microbiology Media
종Listeria
Unit SizeEach