Product Image
Thermo Scientific™
Oxoid™ ONE Broth-Salmonella Supplement
Thermo ScientificTM OxoidTM ONE Broth-Salmonella Supplement is for the selective enrichment of Salmonella spp. from food samples in 24 hours in a single enrichment step.
Have Questions?
Catalog NumberQuantity
SR0242B10/Pk.
Catalog number SR0242B
Price (CLP)
-
Quantity:
10/Pk.
Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Supplement is for the selective enrichment of Salmonella spp. from food samples in 24 hours in a single enrichment step.
  • Add to ONE Broth-Salmonella Part No. CM1091B
  • Each vial supplements 500mL of medium
Specifications
DescriptionONE Broth-Salmonella Supplement
Quantity10/Pk.
YieldEach vial for 225 mL mediμm
FormFreeze-dried
Product TypeMicrobiology Media
SpeciesSalmonella species
Unit SizeEach