Oxoid™ 24 LEB Selective Supplement
Thermo Scientific™

Oxoid™ 24 LEB Selective Supplement

The 24 LEB selective supplement from Oxoid includes ten vials. One vial is required for 500 mL of medium.
Have Questions?
Número do catálogo SR0243E
Preço (BRL)
-
The 24 LEB selective supplement from Oxoid includes ten vials. One vial is required for 500 mL of medium.
  • Selectively enrich Listeria monocytogenes and other Listeria spp. from food and environmental samples
  • Medium provides optimal growing conditions for all Listeria species for PCR applications
  • Specifically designed for use with PCR systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria

Directions:

  • Completely dissolve 21.75 g of 24 LEB base per 500 mL of distilled water
  • Sterilize by autoclaving at 121 °C for 15 min.
  • Cool to 50 °C
  • Aseptically add contents of one vial of 24 LEB selective supplement per 500 mL medium, reconstituted as directed
  • Cool to room temp before use, once made, store medium at 2 to 8 °C in dark

Technique:

  • Refer to BAX™ system user guide and product inserts for PCR Assay L. monocytogenes 24E (QB8125C) and BAX™ system PCR Assay Genus Listeria 24E (QB8135C) for instructions for use
Especificações
AppearanceYellow/orange pellet
Certifications/ComplianceAdheres to ISO 11290-1:1997 standards
For Use With (Application)Selective enrichment of Listeria sp from food and environmental samples
Media TypeDehydrated culture media
Packaging TypeTub
Quantity10 Pack
Shelf Life2 to 5 yr.
Manufacturing OriginUnited Kingdom
SpeciesListeria
Unit SizeEach
Conteúdo e armazenamento
Room Temperature