Oxoid™ Legionella MWY Selective Supplement
Thermo Scientific™

Oxoid™ Legionella MWY Selective Supplement

Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberYield
SR0255BEach vial for 500mL medium
SR0255EEach vial for 100mL medium
Catalog number SR0255B
Price (ZAR)
-
Yield:
Each vial for 500mL medium

Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.

Add to Legionella Agar Base, Part No. CM1203B
Specifications
DescriptionLegionella MWY Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesLegionella
Unit SizeEach