Please note: order placement quantity must be in multiples of 25, minimum of 25 required.
2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, C-Flex 374 MPC insert and body connectors, injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, C-Flex 374 MPC insert and body connectors, injection site

The 20 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aRead more
Have Questions?
Catalog NumberQuantity
SS00022I
also known as SS00022-I
25/Cs.
Catalog number SS00022I
also known as SS00022-I
Price (USD)
-
Quantity:
25/Cs.
The 20 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (one 3/8-inch ID x 5/8-inch OD clear C-Flex 374 (12 in.) MPC insert + cap and pinch clamp, one 3/8-inch ID x 5/8-inch OD clear C-Flex 374 (12 in.) MPC body + cap and pinch clamp, 1 injection site).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Specifications
Capacity (Metric)20 L
Dimensions (L x W x H)63.3 x 42.7 cm (24.9 x 16.8 in.)
Film DescriptionASI™ 28
Includes3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Length (Metric)63.3 cm
Outer PackagingOne 4-mil. poly bag
Port DetailMPC insert and body connectors and injection site
Product LineLabtainer
Quantity25/Cs.
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Tubing StyleC-Flex, EVA
Width (Metric)42.7 cm
MaterialEVA
Product TypeBioprocess Container
Width (English)16.8 in.
Unit Size20 L
Contents & Storage
Store at room temperature.