2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors, injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors, injection site

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)
SS00025I
también denominado SS00025-I
250 mL
SS00024I
también denominado SS00024-I
100 mL
SS00026I
también denominado SS00026-I
500 mL
SS00027I
también denominado SS00027-I
1 L
SS00028I
también denominado SS00028-I
2 L
Número de catálogo SS00025I
también denominado SS00025-I
Precio (USD)
-
Capacidad (métrico):
250 mL

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 2: Luer lock body connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 3: Tubing: EVA, injection port

Especificaciones
Dimensiones16 x 13.5 cm (6.3 x 5.32 in.)
Embalaje exteriorOne 4-mil. poly bag
Línea de productosLabtainer
Cantidad50/Cs.
Condiciones de envíoTemperatura ambiente
Método de esterilizaciónIrradiada
Capacidad (métrico)250 mL
Film DescriptionASI 26/77
MaterialPolietileno
Port DetailLuer Lock Body Connector and Injection Site
Tubing StyleTPE and EVA
Unit Size250 mL
Contenido y almacenamiento
Almacenar a temperatura ambiente.