2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors, injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors, injection site

The 100 mL Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, aWeitere Informationen
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerKapazität (metrisch)
SS00034I
auch als SS00034-I bezeichnet
100 ml
SS00035I
auch als SS00035-I bezeichnet
250 ml
SS00036I
auch als SS00036-I bezeichnet
500 ml
SS00037I
auch als SS00037-I bezeichnet
1 l
SS00038I
auch als SS00038-I bezeichnet
2 l
Katalognummer SS00034I
auch als SS00034-I bezeichnet
Preis (EUR)
-
Kapazität (metrisch):
100 ml
The 100 mL Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film. This BPC includes 3 ports (1 MPC insert + cap, 1 MPC body + cap, 1 injection site).

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: MPC insert with 6.4 mm (1/4 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 2: MPC body with 6.4 mm (1/4 in.) barb, plug, and slide clamp. Tubing: EVA + TPE 374
• Line 3: Tubing: EVA, injection port

Specifications
Abmessungen16 cm x 10 cm (6.3 in. x 3.94 in.)
Außenverpackung4 ml-Polybeutel, ein 4 ml-Polybeutel
ProduktlinieLabtainer
Menge50 per case
VersandbedingungRaumtemperatur
SterilisationsmethodeBestrahlt
Kapazität (metrisch)100 ml
Film DescriptionASI 26/77
MaterialPolyethylen
Port DetailMPC Connector and Injection Port
Tubing StyleTPE and EVA
Unit Size100 mL
Inhalt und Lagerung
Bei Raumtemperatur lagern.