2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert connectors and break-off injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert connectors and break-off injection site

The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aMás información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)
SS00066I
también denominado SS00066-I
100 mL
SS00067I
también denominado SS00067-I
250 mL
SS00068I
también denominado SS00068-I
500 mL
SS00069I
también denominado SS00069-I
1 L
SS00070I
también denominado SS00070-I
2 L
Número de catálogo SS00066I
también denominado SS00066-I
Precio (USD)
-
Capacidad (métrico):
100 mL
The 100 mL Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (2 MPC insert + cap, 1 break-off injection site).

Features of the system include:
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications include:
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquid
• Collecting chromatography feed and fraction
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
• Filters (used for bioburden reduction)—sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers and platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

Especificaciones
Dimensiones12.7 cm x 9.1 cm (5.0 in. x 3.6 in.)
Embalaje exteriorOne 4-mil. poly bag
Línea de productosLabtainer
Cantidad50 per case
ReglamentacióncGMP
Condiciones de envíoTemperatura ambiente
Método de esterilizaciónIrradiada
Capacidad (métrico)100 mL
Film DescriptionASI 28
MaterialEVA
Port DetailMPC insert and body connectors and injection site
Tubing StyleEVA
Unit Size100 mL
Contenido y almacenamiento
Almacenar a temperatura ambiente.