Please note: order placement quantity must be in multiples of 20, minimum of 20 required.
Thermo Scientific™
2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert connectors and injection site
The 10 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aRead more
Have Questions?
Change view
Catalog Number
Capacity (Metric)
SS00074I
also known as SS00074-I
50 L
SS00071I
also known as SS00071-I
5 L
SS00072I
also known as SS00072-I
10 L
SS00073I
also known as SS00073-I
20 L
4 Options
Catalog number SS00074I
also known as SS00074-I
Price (USD)
-
Capacity (Metric):
50 L
The 10 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (2 MPC insert + cap, 1 break-off injection site).
Features of the system include: • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications include: • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters • Preparing and storing buffer, culture media, and process liquid • Collecting chromatography feed and fraction • Harvesting, storing, and transporting bulk drug product and drug precursors
Customize standard BPC systems for optimized single-use technology Choose from the industry’s largest component library, with over 2,000 unique components that include: • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors • Filters (used for bioburden reduction)—sterilization grade filters and vent filters • Tubing—thermoplastic elastomers and platinum-cured silicone • Sample ports—septum-style and needle-free ports • Clamps—sliders, pinch, and crimp bands
Specifications
Dimensions69.3 cm x 58.4 cm (27.3 in. x 23.0 in.)
Outer PackagingOne 4-mil. poly bag
Product LineLabtainer
Quantity20 per case
RegulatorycGMP
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)50 L
Film DescriptionASI 28
MaterialEVA
Port DetailMPC insert connectors and injection site