2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors

The 200 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SS00105I
also known as SS00105-I
200 L
SS00103I
also known as SS00103-I
50 L
SS00104I
also known as SS00104-I
100 L
Catalog number SS00105I
also known as SS00105-I
Price (MXN)
-
Capacity (Metric):
200 L
The 200 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 MPC insert + cap, 1 MPC body + cap).

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: MPC insert with 9.5 mm (3/8 in.) barb, cap, and pinch clamp. Tubing: Silicone, length: 51 cm (20 in.), ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
• Line 2: MPC body with 9.5 mm (3/8 in.) barb, plug, and pinch clamp. Tubing: Silicone, length: 51 cm (20 in.), ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)¬¬¬¬¬¬

Specifications
Capacity (Metric)200 L
Dimensions (L x W x H)142.2 x 95.3 cm (56.0 x 37.5 in.)
Film DescriptionASI™ 26/77
For Use With (Equipment)TK-0100-05
Includes2 Ports (1 MPC Insert + Cap, 1 Mpc Body + Cap)
Length (Metric)142.2 cm
Outer Packaging4 mil. poly bag, One 4-mil. poly bag
Port DetailMPC connections + caps, pinch clamps
Product LineLabtainer
Protective Inner PackagingPoly bags on tubing ends, protective material around rough areas
Quantity200 L, 5 per case
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Tubing StyleC-Flex, 51 cm (20 in.)
Width (Metric)95.3 cm
Length (English)56 in.
MaterialPolyethylene
Product TypeBioprocess Container
SterilitySterile
Width (English)37.5 in.
Unit Size200 L
Contents & Storage
Store at room temperature.