2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Kleenpak insert and body connectors, injection site
2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Kleenpak insert and body connectors, injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Kleenpak insert and body connectors, injection site

The 5 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, aMás información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)
SS00114I
también denominado SS00114-I
20 L
SS00112I
también denominado SS00112-I
5 L
SS00113I
también denominado SS00113-I
10 L
Número de catálogo SS00114I
también denominado SS00114-I
Precio (USD)
-
Capacidad (métrico):
20 L
The 5 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 Kleenpak insert + pinch clamp, 1 Kleenpak body + pinch clamp, 1 injection site).

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: Kleenpak insert and pinch clamp. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 9.5 x 16 mm (0.38 x 0.63 in.)
• Line 2: Kleenpak body and pinch clamp. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 9.5 x 16 mm (0.38 x 0.63 in.)
• Line 3: Injection port

Especificaciones
Dimensiones42.6 cm x 63.5 cm (16.8 in. x 25 in.)
Embalaje exterior4 mil. poly bag, One 4-mil. poly bag
Línea de productosLabtainer
Embalaje interior de protecciónPoly bags on tubing ends, protective material around rough areas
Cantidad25 per case
Condiciones de envíoTemperatura ambiente
Método de esterilizaciónIrradiada
Capacidad (métrico)20 L
Film DescriptionASI 26/77
MaterialPolietileno
Port DetailKleenpak connections and injection port
Tubing StyleTPE, 30 cm (12 in.)
Unit Size20 L
Contenido y almacenamiento
Almacenar a temperatura ambiente.