2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, plugs and injection port
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, plugs and injection port

The 50 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, aMás información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)
SS00122I
también denominado SS00122-I
100 L
SS00121I
también denominado SS00121-I
50 L
SS00123I
también denominado SS00123-I
200 L
Número de catálogo SS00122I
también denominado SS00122-I
Precio (MXN)
-
Capacidad (métrico):
100 L
The 50 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (2 plug + pinch clamp, 1 injection site).

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: Plug 6.4 mm (0.25 in.), pinch clamp. Tubing: C-Flex 374, length: 51 cm (20 in.), ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
• Line 2: Plug 6.4 mm (0.25 in.), pinch clamp. Tubing: C-Flex 374, length: 51 cm (20 in.), ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
• Line 3: Luer injection port. Tubing: C-Flex, length: 7.6 cm (3 in.)

Especificaciones
Dimensiones108 cm x 76.2 cm (42.5 in. x 30 in.)
Para utilizar con (equipo)TK-0100-05
Embalaje exterior4 mil. poly bag, One 4-mil. poly bag
Línea de productosLabtainer
Embalaje interior de protecciónPoly bags on tubing ends, protective material around rough areas
Cantidad10 por caja
Condiciones de envíoTemperatura ambiente
Método de esterilizaciónIrradiada
Capacidad (métrico)100 L
Film DescriptionASI 26/77
MaterialPolietileno
Port Detail1/4 in. Plug and Injection Port
Tubing StyleC-Flex, 51 cm (20 in.)
Unit Size100 L
Contenido y almacenamiento
Almacenar a temperatura ambiente.