Thermo Scientific™
Smart Probes with Calibration/Set-up Memory
Catalog number: SSPA3
Thermo Scientific™
Smart Probes with Calibration/Set-up Memory
Catalog number: SSPA3
The “Smart” configuration for Thermo Scientific™ probes includes a memory device containing all pertinent calibration and operating criteria; boosts efficiency and reduces user workload.
Connection of a smart probe to a compatible instrument means a high degree of automation and efficiency.
Catalog Number
SSPA3
Unit Size
Each
Description
Gamma Nal(TI) scintillation probe
Price (USD)
Full specifications
TypeSmart Probe
DescriptionGamma Nal(TI) scintillation probe
Unit SizeEach
Catalog Number
Unit Size
Each
Description
End window GM probe
Price (USD)
Full specifications
TypeSmart Probe
DescriptionEnd window GM probe
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Pancake GM probe with aluminum shield
Price (USD)
Full specifications
TypeSmart Probe
DescriptionPancake GM probe with aluminum shield
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Pancake GM probe with tungsten shield
Price (USD)
Full specifications
TypeSmart Probe
DescriptionPancake GM probe with tungsten shield
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Beta-Gamma GM probe, energy compensated
Price (USD)
Full specifications
TypeSmart Probe
DescriptionBeta-Gamma GM probe, energy compensated
Unit SizeEach
Catalog Number
Unit Size
Each
Description
High-range GM dose rate probe
Price (USD)
Full specifications
TypeSmart Probe
DescriptionHigh-range GM dose rate probe
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Pancake GM probe
Price (USD)
Full specifications
TypeSmart Probe
DescriptionPancake GM probe
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Alpha/Beta scintillation, Smart probe
Price (USD)
Full specifications
TypeSmart Probe
DescriptionAlpha/Beta scintillation, Smart probe
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Alpha scintillation smart probe
Price (USD)
Full specifications
TypeSmart Probe
DescriptionAlpha scintillation smart probe
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Neutron rem detector ball
Price (USD)
Full specifications
TypeSmart Probe
DescriptionNeutron rem detector ball
Unit SizeEach
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
SSPA3Full specifications
EachGamma Nal(TI) scintillation probeRequest A Quote
TypeSmart Probe
DescriptionGamma Nal(TI) scintillation probe
Unit SizeEach
SHP190AFull specifications
EachEnd window GM probeRequest A Quote
TypeSmart Probe
DescriptionEnd window GM probe
Unit SizeEach
SHP210ALFull specifications
EachPancake GM probe with aluminum shieldRequest A Quote
TypeSmart Probe
DescriptionPancake GM probe with aluminum shield
Unit SizeEach
SHP210TFull specifications
EachPancake GM probe with tungsten shieldRequest A Quote
TypeSmart Probe
DescriptionPancake GM probe with tungsten shield
Unit SizeEach
SHP270Full specifications
EachBeta-Gamma GM probe, energy compensatedRequest A Quote
TypeSmart Probe
DescriptionBeta-Gamma GM probe, energy compensated
Unit SizeEach
SHP290Full specifications
EachHigh-range GM dose rate probeRequest A Quote
TypeSmart Probe
DescriptionHigh-range GM dose rate probe
Unit SizeEach
SHP360Full specifications
EachPancake GM probeRequest A Quote
TypeSmart Probe
DescriptionPancake GM probe
Unit SizeEach
SHP380ABFull specifications
EachAlpha/Beta scintillation, Smart probeRequest A Quote
TypeSmart Probe
DescriptionAlpha/Beta scintillation, Smart probe
Unit SizeEach
SHP380AFull specifications
EachAlpha scintillation smart probeRequest A Quote
TypeSmart Probe
DescriptionAlpha scintillation smart probe
Unit SizeEach
SNRDFull specifications
EachNeutron rem detector ballRequest A Quote
TypeSmart Probe
DescriptionNeutron rem detector ball
Unit SizeEach
Showing 10 of 11Show more
  • Automatically sets the high voltage, threshold settings, units of measurement, response times, over range limits and other defining characteristics
  • Permits rapid detector replacements or swapping of probe types in the field
  • Facilitates a more efficient means for calibrating instruments and probes
Ordering Information:
Smart probes may be ordered by using the prefix “S” to any HP probe model number, i.e. SHP-270. Existing probes may also be converted to the Smart configuration by the addition of an external SmartPack