Smart-Tracker™ Wireless Data Module Accessories
Actual product may vary
Smart-Tracker™ Wireless Data Module Accessories
Smart-Tracker™ Wireless Data Module Accessories
Thermo Scientific™
Smart-Tracker™ Wireless Data Module Accessories
Maximize Thermo Scientific™ Smart-Tracker™ with additional accessories or personalize your Smart-Tracker with a series of colored bands.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
ST501-5055 colored rings, purple
ST202-200Temperature sensor -200/+200°C
ST500-500Battery VARTA 1200mA (pack 4)
ST500-501Battery VARTA 600mA (pack 4)
ST501-5005 colored rings, gray
ST501-5015 colored rings, ocean blue
ST501-5025 colored rings, green
ST501-5035 colored rings, gold
ST501-504
ST501-5065 colored rings, black
ST501-5075 colored rings, yellow
ST501-5085 colored rings, sky blue
ST501-509Set of 10 rings (all colors)
ST502-50050 stickers QR codes
Catalog number ST501-505
Price (MXN)
-
Description:
5 colored rings, purple
Maximize Thermo Scientific™ Smart-Tracker™ with additional accessories or personalize your Smart-Tracker with a series of colored bands.
  • Warranty varies by country, please contact us for details
  • Specifications
    Description5 colored rings, purple
    Dimensions (L x W x H)0.24 x 0.63 x 1.02 in. (6 x 16 x 26 mm) [Shipping Dimensions]
    For Use With (Application)Wireless Monitoring Solution
    For Use With (Equipment)Smart-Tracker™ Module
    Shipping Weight (English)0.33 lb.
    Shipping Weight (Metric)0.15 kg
    TypeRings
    Unit SizeEach