Oxoid™ Antimicrobial Susceptibility disc Dispenser
Oxoid™ Antimicrobial Susceptibility disc Dispenser
Oxoid™ Antimicrobial Susceptibility disc Dispenser
Oxoid™ Antimicrobial Susceptibility disc Dispenser
Thermo Scientific™
Oxoid™ Antimicrobial Susceptibility disc Dispenser
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoNo. of PlacesFor Use With (Equipment)
ST61006 Oxoid Disc Cartridges100mm diameter plates
ST60906 Oxoid Disc Cartridges90mm diameter plates
ST80908 Oxoid Disc Cartridges90mm diameter plates
ST81008 Oxoid Disc Cartridges100mm diameter plates
Número do catálogo ST6100
Preço (BRL)
-
No. of Places:
6 Oxoid Disc Cartridges
For Use With (Equipment):
100mm diameter plates
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-tamping Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility disc Dispenser.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For use only with Oxoid™ Antimicrobial Susceptibility discs.
Especificações
Certifications/ComplianceFor use only with Oxoid Antimicrobial Susceptibility Discs
DescriptionOxoid Antimicrobial Susceptibility Disc Dispenser
For Use With (Equipment)100mm diameter plates
No. of Places6 Oxoid Disc Cartridges
Packaging Type1 Dispenser/Pk.
Product TypeAntimicrobial Susceptibility Disc Dispenser
Unit SizeEach