Orion™ Universal Power Adapter for Orion™ Versa Star Meters
Thermo Scientific™
Orion™ Universal Power Adapter for Orion™ Versa Star Meters
Ensure a constant power supply to your meter using a spare or replacement Thermo Scientific™ Orion™ Universal Power Adapter for the Thermo Scientific™ Orion™ Versa Star meters
Have Questions?
Catalog number STARA-PWR
Price (USD)
82.89
Online Exclusive
92.10
Save 9.21 (10%)
Each
Price (USD)
82.89
Online Exclusive
92.10
Save 9.21 (10%)
Each
Ensure a constant power supply to your meter using a spare or replacement Thermo Scientific™ Orion™ Universal Power Adapter for the Thermo Scientific™ Orion™ Versa Star meters.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionOrion Versa Star universal power adapter
Electrical Requirements100/240 VAC, 50/60 Hz
Frequency50/60 Hz
IncludesUniversal power adapter with US, EU, UK and China plug plates
Voltage100/240 VAC
TypeMeter Accessories
Unit SizeEach