Thermo Scientific™
Orion Star™ A111 Benchtop pH Meter
Catalog number: STARA1117
Thermo Scientific™
Orion Star™ A111 Benchtop pH Meter
Catalog number: STARA1117
Take simple, routine pH, mV and temperature measurements with the Thermo Scientific™ Orion Star™ A111 Benchtop pH Meter. This meter is an easy-to-use and budget-friendly instrument for basic pH analysis in the lab. Perform up to a 3 point pH calibration with easy recall of calibration points and slope. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
Catalog Number
STARA1117
Unit Size
Each
Description
Orion Star A111 benchtop pH meter TRIS kit with TRIS-compatible ROSS Ultra pH/ATC electrode
Includes
Star A111 pH meter, 8157BNUMD ROSS Ultra epoxy-body pH/ATC electrode, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter
Price (USD)
1,070.00
Your Price:
Availability
***
Full specifications
Quantity
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Battery Life2000 hr.
Battery Type4 x AA (sold separately)
Certifications/ComplianceCE, TUV 3-1, FCC Class A
Channels1
DescriptionOrion Star A111 benchtop pH meter TRIS kit with TRIS-compatible ROSS Ultra pH/ATC electrode
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.01
Accuracy (Temperature)±0.1°C
CalibrationpH, relative mV (RmV) and temperature
Calibration Points1 to 3
Data Management DetailsInternal data point memory
Data Points50
Display TypeLCD
Log Function TypeAutomatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
Measurement ModespH, mV or relative mV (RmV) with temperature
OutputsNone
Range (mV)±1600.0 mV
Range (pH)-2 to 16
Resolution (pH)0.1, 0.01
Temperature Range (Metric)-5°C to +105°C
IncludesStar A111 pH meter, 8157BNUMD ROSS Ultra epoxy-body pH/ATC electrode, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
IP RatingIP-54
Isopotential Point7.00pH
KeypadSimple with highlighted measure key, scroll and shortcut keys
LanguagesEnglish
MemoryNon-volatile memory preserves log and settings
ModelOrion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power SupplyUniversal AC adapter (included) or 4 AA batteries (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (includes 8157BNUMD ROSS Ultra Triode pH/ATC electrode)
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (mV)0.1mV
Resolution (Temperature)0.1°C, 0.1°F
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 Year Meter Warranty
Dimensions (L x W x H)24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
Weight (English)2 lb.
Weight (Metric)0.9 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Orion Star A111 benchtop pH meter with stand
Includes
Star A111 pH meter, electrode stand, 100-240V universal power adapter
Price (USD)
600.00
Your Price:
Availability
***
Full specifications
Quantity
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.01
CalibrationpH, relative mV (RmV) and temperature
Calibration Points1 point relative mV (RmV) mode
Data Management DetailsInternal data point memory
Data Points50
Display TypeLCD
Log Function TypeAutomatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
Measurement ModespH, mV or relative mV (RmV) with temperature
OutputsNone
Temperature Range (Metric)-5°C to +105°C
Battery Life2000 hr.
Battery Type4 x AA (sold separately)
Channels1
Certifications/ComplianceCE, TUV 3-1, FCC Class A
DescriptionOrion Star A111 benchtop pH meter with stand
Dimensions (L x W x H)24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
IncludesStar A111 pH meter, electrode stand, 100-240V universal power adapter
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
IP RatingIP-54
Isopotential Point7.00pH
KeypadSimple with highlighted measure key, scroll and shortcut keys
LanguagesEnglish
MemoryNon-volatile memory preserves log and settings
ModelOrion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power SupplyUniversal AC adapter (included) or 4 AA batteries (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (purchase separately)
Range (mV)±1600.0 mV
Range (pH)-2 to 16
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (pH)0.1, 0.01
Resolution (mV)0.1mV
Resolution (Temperature)0.1°C, 0.1°F
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 Year Meter Warranty
Weight (English)2 lb.
Weight (Metric)0.9 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Orion Star A111 benchtop pH meter standard kit with general purpose pH/ATC electrode
Includes
Star A111 pH meter, 9157BNMD epoxy-body pH/ATC electrode, 916099 pH buffer kit, electrode stand, 100-240V universal power adapter
Price (USD)
888.00
Your Price:
Availability
***
Full specifications
Quantity
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.01
CalibrationpH, relative mV (RmV) and temperature
Calibration Points1 to 3
Data Management DetailsInternal data point memory
Data Points50
Display TypeLCD
Log Function TypeAutomatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
Measurement ModespH, mV or relative mV (RmV) with temperature
OutputsNone
Temperature Range (Metric)-5°C to +105°C
Battery Life2000 hr.
Battery Type4 x AA (sold separately)
Channels1
Certifications/ComplianceCE, TUV 3-1, FCC Class A
DescriptionOrion Star A111 benchtop pH meter standard kit with general purpose pH/ATC electrode
Dimensions (L x W x H)24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
IncludesStar A111 pH meter, 9157BNMD epoxy-body pH/ATC electrode, 916099 pH buffer kit, electrode stand, 100-240V universal power adapter
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
IP RatingIP-54
Isopotential Point7.00pH
KeypadSimple with highlighted measure key, scroll and shortcut keys
LanguagesEnglish
MemoryNon-volatile memory preserves log and settings
ModelOrion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power SupplyUniversal AC adapter (included) or 4 AA batteries (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (includes 9157BNMD Orion Triode pH/ATC electrode)
Range (mV)±1600.0 mV
Range (pH)-2 to 16
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (pH)0.1, 0.01
Resolution (mV)0.1mV
Resolution (Temperature)0.1°C, 0.1°F
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 Year Meter Warranty
Weight (English)2 lb.
Weight (Metric)0.9 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Orion Star A111 benchtop pH meter bio kit with TRIS-compatible double junction pH electrode and ATC probe
Includes
Star A111 pH meter, GD9156BNWP Double Junction pH electrode, 927007MD stainless steel ATC probe, electrode stand, 100-240V universal power adapter
Price (USD)
951.00
Your Price:
Availability
***
Full specifications
Quantity
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.01
CalibrationpH, relative mV (RmV) and temperature
Calibration Points1 to 3
Data Management DetailsInternal data point memory
Data Points50
Display TypeLCD
Log Function TypeAutomatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
Measurement ModespH, mV or relative mV (RmV) with temperature
OutputsNone
Temperature Range (Metric)-5°C to +105°C
Battery Life2000 hr.
Battery Type4 x AA (sold separately)
Channels1
Certifications/ComplianceCE, TUV 3-1, FCC Class A
DescriptionOrion Star A111 benchtop pH meter bio kit with TRIS-compatible double junction pH electrode and ATC probe
Dimensions (L x W x H)24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
IncludesStar A111 pH meter, GD9156BNWP Double Junction pH electrode, 927007MD stainless steel ATC probe, electrode stand, 100-240V universal power adapter
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
IP RatingIP-54
Isopotential Point7.00pH
KeypadSimple with highlighted measure key, scroll and shortcut keys
LanguagesEnglish
MemoryNon-volatile memory preserves log and settings
ModelOrion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power SupplyUniversal AC adapter (included) or 4 AA batteries (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (includes GD9156BNWP Double Junction pH electrode, 927007MD Orion ATC probe)
Range (mV)±1600.0 mV
Range (pH)-2 to 16
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (pH)0.1, 0.01
Resolution (mV)0.1mV
Resolution (Temperature)0.1°C, 0.1°F
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 Year Meter Warranty
Weight (English)2 lb.
Weight (Metric)0.9 kg
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeDescriptionIncludesPrice (USD)AvailabilityQuantity
STARA1117Full specifications
EachOrion Star A111 benchtop pH meter TRIS kit with TRIS-compatible ROSS Ultra pH/ATC electrodeStar A111 pH meter, 8157BNUMD ROSS Ultra epoxy-body pH/ATC electrode, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter
1,070.00
Your Price:
***
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Battery Life2000 hr.
Battery Type4 x AA (sold separately)
Certifications/ComplianceCE, TUV 3-1, FCC Class A
Channels1
DescriptionOrion Star A111 benchtop pH meter TRIS kit with TRIS-compatible ROSS Ultra pH/ATC electrode
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.01
Accuracy (Temperature)±0.1°C
CalibrationpH, relative mV (RmV) and temperature
Calibration Points1 to 3
Data Management DetailsInternal data point memory
Data Points50
Display TypeLCD
Log Function TypeAutomatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
Measurement ModespH, mV or relative mV (RmV) with temperature
OutputsNone
Range (mV)±1600.0 mV
Range (pH)-2 to 16
Resolution (pH)0.1, 0.01
Temperature Range (Metric)-5°C to +105°C
IncludesStar A111 pH meter, 8157BNUMD ROSS Ultra epoxy-body pH/ATC electrode, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
IP RatingIP-54
Isopotential Point7.00pH
KeypadSimple with highlighted measure key, scroll and shortcut keys
LanguagesEnglish
MemoryNon-volatile memory preserves log and settings
ModelOrion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power SupplyUniversal AC adapter (included) or 4 AA batteries (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (includes 8157BNUMD ROSS Ultra Triode pH/ATC electrode)
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (mV)0.1mV
Resolution (Temperature)0.1°C, 0.1°F
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 Year Meter Warranty
Dimensions (L x W x H)24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
Weight (English)2 lb.
Weight (Metric)0.9 kg
Unit SizeEach
STARA1110Full specifications
EachOrion Star A111 benchtop pH meter with standStar A111 pH meter, electrode stand, 100-240V universal power adapter
600.00
Your Price:
***
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.01
CalibrationpH, relative mV (RmV) and temperature
Calibration Points1 point relative mV (RmV) mode
Data Management DetailsInternal data point memory
Data Points50
Display TypeLCD
Log Function TypeAutomatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
Measurement ModespH, mV or relative mV (RmV) with temperature
OutputsNone
Temperature Range (Metric)-5°C to +105°C
Battery Life2000 hr.
Battery Type4 x AA (sold separately)
Channels1
Certifications/ComplianceCE, TUV 3-1, FCC Class A
DescriptionOrion Star A111 benchtop pH meter with stand
Dimensions (L x W x H)24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
IncludesStar A111 pH meter, electrode stand, 100-240V universal power adapter
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
IP RatingIP-54
Isopotential Point7.00pH
KeypadSimple with highlighted measure key, scroll and shortcut keys
LanguagesEnglish
MemoryNon-volatile memory preserves log and settings
ModelOrion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power SupplyUniversal AC adapter (included) or 4 AA batteries (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (purchase separately)
Range (mV)±1600.0 mV
Range (pH)-2 to 16
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (pH)0.1, 0.01
Resolution (mV)0.1mV
Resolution (Temperature)0.1°C, 0.1°F
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 Year Meter Warranty
Weight (English)2 lb.
Weight (Metric)0.9 kg
Unit SizeEach
STARA1115Full specifications
EachOrion Star A111 benchtop pH meter standard kit with general purpose pH/ATC electrodeStar A111 pH meter, 9157BNMD epoxy-body pH/ATC electrode, 916099 pH buffer kit, electrode stand, 100-240V universal power adapter
888.00
Your Price:
***
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.01
CalibrationpH, relative mV (RmV) and temperature
Calibration Points1 to 3
Data Management DetailsInternal data point memory
Data Points50
Display TypeLCD
Log Function TypeAutomatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
Measurement ModespH, mV or relative mV (RmV) with temperature
OutputsNone
Temperature Range (Metric)-5°C to +105°C
Battery Life2000 hr.
Battery Type4 x AA (sold separately)
Channels1
Certifications/ComplianceCE, TUV 3-1, FCC Class A
DescriptionOrion Star A111 benchtop pH meter standard kit with general purpose pH/ATC electrode
Dimensions (L x W x H)24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
IncludesStar A111 pH meter, 9157BNMD epoxy-body pH/ATC electrode, 916099 pH buffer kit, electrode stand, 100-240V universal power adapter
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
IP RatingIP-54
Isopotential Point7.00pH
KeypadSimple with highlighted measure key, scroll and shortcut keys
LanguagesEnglish
MemoryNon-volatile memory preserves log and settings
ModelOrion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power SupplyUniversal AC adapter (included) or 4 AA batteries (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (includes 9157BNMD Orion Triode pH/ATC electrode)
Range (mV)±1600.0 mV
Range (pH)-2 to 16
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (pH)0.1, 0.01
Resolution (mV)0.1mV
Resolution (Temperature)0.1°C, 0.1°F
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 Year Meter Warranty
Weight (English)2 lb.
Weight (Metric)0.9 kg
Unit SizeEach
STARA1116Full specifications
EachOrion Star A111 benchtop pH meter bio kit with TRIS-compatible double junction pH electrode and ATC probeStar A111 pH meter, GD9156BNWP Double Junction pH electrode, 927007MD stainless steel ATC probe, electrode stand, 100-240V universal power adapter
951.00
Your Price:
***
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.01
CalibrationpH, relative mV (RmV) and temperature
Calibration Points1 to 3
Data Management DetailsInternal data point memory
Data Points50
Display TypeLCD
Log Function TypeAutomatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
Measurement ModespH, mV or relative mV (RmV) with temperature
OutputsNone
Temperature Range (Metric)-5°C to +105°C
Battery Life2000 hr.
Battery Type4 x AA (sold separately)
Channels1
Certifications/ComplianceCE, TUV 3-1, FCC Class A
DescriptionOrion Star A111 benchtop pH meter bio kit with TRIS-compatible double junction pH electrode and ATC probe
Dimensions (L x W x H)24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
IncludesStar A111 pH meter, GD9156BNWP Double Junction pH electrode, 927007MD stainless steel ATC probe, electrode stand, 100-240V universal power adapter
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
IP RatingIP-54
Isopotential Point7.00pH
KeypadSimple with highlighted measure key, scroll and shortcut keys
LanguagesEnglish
MemoryNon-volatile memory preserves log and settings
ModelOrion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power SupplyUniversal AC adapter (included) or 4 AA batteries (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (includes GD9156BNWP Double Junction pH electrode, 927007MD Orion ATC probe)
Range (mV)±1600.0 mV
Range (pH)-2 to 16
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (pH)0.1, 0.01
Resolution (mV)0.1mV
Resolution (Temperature)0.1°C, 0.1°F
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 Year Meter Warranty
Weight (English)2 lb.
Weight (Metric)0.9 kg
Unit SizeEach
Showing 4 of 4
Features:
  • Large, informative screen clearly shows the main information, including pH or mV reading, temperature, meter mode, calibration information and power source.
  • Thermo Scientific™ AUTO-READ™ locks in the stable reading on your screen and ready indicator alerts when readings are stable.
  • Meter offers up to 3 point pH calibration and automatic recognition of USA/NIST and DIN buffers.
  • Use can easily recall calibration slope data for procedure checks.
  • Nonvolatile memory holds up to 50 data points.
  • Included electrode arm and newly designed probe holder make it easier to maintain and place probes into samples.
  • Plug the meter into almost any power outlet with the included universal power adapter or use four AA batteries (sold separately) to run the meter on DC power.
  • The IP54-rated housing can handle splashes and can be wall-mounted if table space is a problem.
Includes:
Orion STAR: A111 Benchtop pH Meter, Part no. STARA1110, includes electrode arm with redesigned holder and universal power adapter
Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1115, includes 9157BNUMD Orion Triode™ 3-in-1 pH/ATC probe; Solution kit (60mL each of pH 4, 7 and 10 buffers; storage solution and cleaning solution); electrode arm with redesigned holder and universal power adapter
Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1116, includes GD9156BNWP Orion Green electrode; 927007MD Stainless Steel ATC Probe; electrode arm with redesigned holder and universal power adapter
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Figures