Thermo Scientific™
Lindberg/Blue M™ 1500°C General-Purpose Tube Furnaces
 Related applications:
Lab Ovens Furnaces
Featuring integral control designed for a range of applications which require processing flexibility with fast heatup and recovery. Thermo Scientific™ Lindberg/Blue M 1500°C General-Purpose Tube Furnaces incorporate Moldatherm™ insulation to increase temperature uniformity, improve energy efficiency and to maintain low exterior cabinet temperatures during operation.
Not available in Europe
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Accommodate 1in., 2in. and 3in. O.D. process tubes (customer supplied)
  • Silicon carbide heating elements positioned above and below tube works with Type “R” thermocouple to stabilize temperature
  • Microprocessor-based PID programmable control (proportional, integral, derivative) prevents overshoot
  • Available with Single Program/Multiple Segment or Multiple Program/Multiple Segment Programmable Controller
  • Program(s) and segment(s) for ramp (up and down) and dwell (timed hold) temperature control
  • Adjustable high limit overtemperature protection
  • Simultaneous LED display of actual temperature vs. setpoint
  • Temperature display in °C or °F
  • Optional Overtemperature Control (OTC) on Multi-Program/Multi-Segment model
Documents
Manuals & protocols