Sensititre™ STP6F
Thermo Scientific™
Sensititre™ STP6F
Perform susceptibility testing of Streptococcus species with this FDA cleared, convenient plate including Tigecycline.
Have Questions?
產品號碼Quantity
STP6F10/Pk.
產品號碼 STP6F
價格 (TWD)
-
Quantity:
10/Pk.
Perform susceptibility testing of Streptococcus species with Thermo Scientific™ Sensititre™ Streptococcus STP6F AST Plate. This plate is FDA-cleared for automated or manual reading of Streptococcus species using the Thermo Scientific™ Sensititre™ ARIS™ 2X ID/AST System, Thermo Scientific™ Sensititre™ OptiRead System, or Sensititre™ manual platforms, including the Thermo Scientific™ Sensititre™ Vizion™ System for maximum versatility.
Eliminate offline tests with a full range of antimicrobials, including Tigecycline, with the Sensititre Streptococcus STP6F AST Plate. 
  • Individual packaging allows laboratories to test one isolate at a time
  • Test 20 antimicrobials on a single AST plate for cost-effective results
  • True MIC results provide better resistance tracking
  • Extended shelf life and room-temperature storage eliminate inventory control concerns
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
規格
0.5 McFarland StandardMHB (Part No. T346205)
Antibiotic CodeVarious
Antimicrobial AgentsVarious
Broth TypeMHB w/ LHB (Part No. CP114)
DescriptionSensititre Streptococcus species MIC Plate
For Use With (Application)All Streptococcus sp.
Incubation Conditions34-36°C O2
Incubation Time20-24 hr
Isolates Per Plate1
No. per Pack10
Packaging TypeBox
Quantity10/Pk.
Reading OptionsAutoread or manual
Transfer Volume100µL
Volume Per Well100 μL
Product TypeAST Plate
SpeciesStreptococcus species
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach