Product Image
Thermo Scientific™

Thermo Scientific™ 5:1 Single-Use Bioreactor, jacketed, AC motor, 240VAC, e-box, analog load cells, 1000 L

This 1000 L Thermo Scientific 5:1 Single-Use Bioreactor (SUB) is water-jacketed and includes AC motor, 240VAC, e-box, and analog loadRead more
Have Questions?
Catalog NumberQuantity
SUB100090101,000 L
Catalog number SUB10009010
Price (EUR)
-
Quantity:
1,000 L
This 1000 L Thermo Scientific 5:1 Single-Use Bioreactor (SUB) is water-jacketed and includes AC motor, 240VAC, e-box, and analog load cells. It is designed using traditional stainless-steel bioreactor principles to ensure optimal cell culture performance. The SUB is a fully functional bioreactor vessel including single-use sterile contact surfaces for mixing, venting, sparging, and temperature sensing. The complete line of SUBs include 50, 100, 250, 500, 1000, and 2000-L sizes that ensure consistent scalability from pilot-scale studies, pre-clinical to commercial production.

Features:
• Unique proprietary mixing system that enables conventional overhead mixing while maintaining sterility and integrity of the SUB BioProcess Container (BPC)
• Largest qualified standard component library that allows for customization of SUB BPCs
• Minimized vessel footprint and reduced hold-up volumes
• Offered with AC or DC motors designed for precise agitation control
• Optional cable management tree provides line set and cabling organization for ease of use
• Load cells offered as standard feature with analog or digital communication options
• Option to use industry-leading single-use and traditional sensors for pH, DO, and pressure monitoring
• Option to choose the bioreactor vessel as a stand-alone system or complete turnkey system with dedicated controls from either the Finesse™ or Applikon™ families of control systems for bioproduction. These systems work on PC, Delta V™, Allen-Bradley™, or Siemens™ software platforms
• Updated 5:1 mixing turn-down ratio in combination with patented cross-flow sparger technology introduces gas just above liquid’s surface to facilitate efficient mass transfer CO2 stripping and reduce bubble sparging and associated foam damage to cells. This feature allows customers to do more with fewer vessels, which improves productivity and reduces costs simultaneously.

Applications of the Thermo Scientific SUB include:
• Batch, fed batch, and perfusion cultures
• Suspension and microcarrier cultures
• Proven for various cell lines such as CHO, Sp2/0 hybridoma, PER.C6, HEK293, Vero, MDCK, etc.

Accessories:
• Tube Assembly, Cat # SH3072002
• Molded Probe Holder, Cat # SV50177P01

Specifications
Product LineHyPerforma
Quantity1,000 L
Capacity (Metric)1000 L
MaterialStainless Steel
Unit Size1,000 L
Contents & Storage
Store at room temperature.