HyPerforma™ Mixtainer™ Hardware, Magnetic Stirring Plate; US 115V, 50/60 Hz, 1 amp
HyPerforma™ Mixtainer™ Hardware, Magnetic Stirring Plate; US 115V, 50/60 Hz, 1 amp
Thermo Scientific™
HyPerforma™ Mixtainer™ Hardware, Magnetic Stirring Plate; US 115V, 50/60 Hz, 1 amp
Have Questions?
Catalog NumberQuantity
SV30097021 Ea.
Catalog number SV3009702
Price (USD)
-
Request A Quote
Quantity:
1 Ea.
The Thermo Scientific™ Magnetic Stirring Plate (US 115V, 50/60 Hz, 1 amp) is part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications. Additional components of the system include the Thermo Scientific™ HyPerforma™ Mixtainer™ BioProcess Container (BPC) with an integrated magnetic stir bar, a conical drum, a support plate and a dolly. The optimized support hardware allows mixing, moving, storage, shipping and discharge of the BPC contents while maintaining sterility of the contents. Standard products are available for sterile liquid-to-liquid or powder-to-liquid mixing.
Recommended for:
  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions
Specifications
Power Requirements115V 50/60Hz; 1A
Product LineHyPerforma, Mixtainer
Quantity1 Ea.
Voltage115 V
Unit Size1 each