16EZA/16EZB Holders, 304 SS, Mechanical Polish for 3M Harvest RC Chromatographic Clarifier
16EZA/16EZB Holders, 304 SS, Mechanical Polish for 3M Harvest RC Chromatographic Clarifier
Thermo Scientific™

16EZA/16EZB Holders, 304 SS, Mechanical Polish for 3M Harvest RC Chromatographic Clarifier

This innovative solution is a harvest and clarification system for Chinese hamster ovary (CHO) derived cell cultures. The 3M Harvest RC Chromatographic Clarifier enables single-stage clarification of low- to high-density CHO cell cultures with high product recovery while reducing consumption of buffer and water and simplifying scale-up from laboratory to manufacturing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)Holds
SV30208.04210.7 x 108.7 x 108.7 cm7
SV30208.01121 x 65.6 x 65.6 cm1
SV30208.02148 x 65.6 x 65.6 cm2
SV30208.03169 x 65.6 x 65.6 cm3
Catalog number SV30208.04
Price (MXN)
-
Request A Quote
Dimensions (L x W x H):
210.7 x 108.7 x 108.7 cm
Holds:
7

Thermo Fisher Scientific is an authorized reseller of the 3M Harvest RC Chromatographic Clarifier, which is a harvest and clarification system for Chinese hamster ovary (CHO)-derived cell cultures. By employing synthetic fibrous AEX chromatographic clarification media, the 3M Harvest RC clarifier enables single-stage clarification of low- to high-density CHO cell cultures (>40 million cells per mL) with high product recovery. This novel method of harvest clarification condenses the multiple stages of traditional process into a single stage while reducing consumption of buffer and water compared to depth filtration. Predictable scalability from laboratory to manufacturing capsules allows for simplified process development and planning. The 3M Harvest RC clarifier is suitable for your harvest needs from R&D through clinical trials and manufacturing. The manufacturing-scale harvest solution requires a stainless-steel holder as well as single-use cartridges, while the R&D and pilot-scale systems require only single-use cartridges.

  • Simplification of high-cell-density cell-culture-fluid clarification-unit operations
  • Optimized for PCV high density CHO cell culture (5–8% PCV)
  • Replacement of primary, secondary, and guard membrane clarification stages
  • Typical product recovery of 95+% (capsules)
  • Synthetic chromatographic harvest media with chemically defined extractables
  • Predictable scaling from discovery to manufacturing
  • Lower total cost of manufacturing compared to centrifugation and depth filtration
  • Lower consumption of buffer and water compared to depth filtration
  • Capsules fit into laboratory to manufacturing scale workflows
Specifications
Dimensions (L x W x H)210.7 x 108.7 x 108.7 cm
Holds7
TypeLarge Production Holder
MaterialStainless Steel (304)
Unit SizeEach