3M™ Harvest RC BC Capsules
3M™ Harvest RC BC Capsules
Thermo Scientific™

3M™ Harvest RC BC Capsules

This innovative solution is a harvest and clarification system for Chinese hamster ovary (CHO) derived cell cultures. The 3M Harvest RC Chromatographic Clarifier enables single-stage clarification of low- to high-density CHO cell cultures with high product recovery while reducing consumption of buffer and water and simplifying scale-up from laboratory to manufacturing.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume (Metric)
SV30208.15160 L
SV30208.1423 L
Número do catálogo SV30208.15
Preço (BRL)
-
Requisite uma Cotação
Volume (Metric):
160 L

Thermo Fisher Scientific is an authorized reseller of the 3M Harvest RC Chromatographic Clarifier, which is a harvest and clarification system for Chinese hamster ovary (CHO)-derived cell cultures. By employing synthetic fibrous AEX chromatographic clarification media, the 3M Harvest RC clarifier enables single-stage clarification of low- to high-density CHO cell cultures (>40 million cells per mL) with high product recovery. This novel method of harvest clarification condenses the multiple stages of traditional process into a single stage while reducing consumption of buffer and water compared to depth filtration. Predictable scalability from laboratory to manufacturing capsules allows for simplified process development and planning. The 3M Harvest RC clarifier is suitable for your harvest needs from R&D through clinical trials and manufacturing. The manufacturing-scale harvest solution requires a stainless-steel holder as well as single-use cartridges, while the R&D and pilot-scale systems require only single-use cartridges.

  • Simplification of high-cell-density cell-culture-fluid clarification-unit operations
  • Optimized for PCV high density CHO cell culture (5–8% PCV)
  • Replacement of primary, secondary, and guard membrane clarification stages
  • Typical product recovery of 95+% (capsules)
  • Synthetic chromatographic harvest media with chemically defined extractables
  • Predictable scaling from discovery to manufacturing
  • Lower total cost of manufacturing compared to centrifugation and depth filtration
  • Lower consumption of buffer and water compared to depth filtration
  • Capsules fit into laboratory to manufacturing scale workflows
Especificações
AutoclavableNot Autoclavable
Diameter (Metric)45.2 cm
Filtration Area1.61 m2
For Use With (Application)Biopharmaceutical Manufacturing, Bioprocessing, Cell Culture Clarification, DNA Reduction, Fiber Chromatography, mAb Production, Single Stage Clarification, Turbidity Reduction
Height (Metric)20.3 cm
Individually wrappedYes
Inlet Connection (English)1 1/2 in.
Inlet StyleSanitary Connector
Max. Pressure50 psig
ModelHarvest RC BC16000
No. per Case1
No. per Pack1
Outlet Fitting1 1/2 in.
Outlet StyleSanitary Connector
Temperature (English) Max.104 °F
Temperature (English) Operating32°F to 104°F
Temperature (Metric) Max40 °C
Temperature (Metric) Operating0°C to 40°C
Temperature Range (English)32°F to 104°F
Temperature Range (Metric)0°C to 40°C
TypeCapsule
Volume (Metric)160 L
Weight (Metric)9.8 kg
DisposableSingle-use
For Use With (Equipment)SV30208.01/.02/.03/04, SV30208.05/.06
MaterialFluorocarbon, Glass-filled Polypropylene, Polycarbonate, Polypropylene, Polysulfone, Silicone, TPE
Material (Membrane)PES
Pore Size0.2 μm
SterilityNon-sterile
Unit SizeEach