Thermo Scientific™
HyPerforma™ Smartainer™ BPC Hoist, 1500L Smartainer System
Catalog number: SV50161.22
Have Questions?
Thermo Scientific™
HyPerforma™ Smartainer™ BPC Hoist, 1500L Smartainer System
Catalog number: SV50161.22
Have Questions?
Catalog Number
SV50161.22
Unit Size
Price (USD)
Catalog NumberUnit SizePrice (USD)
SV50161.22Request A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific™ HyPerforma™ BPC Hoist assists in handling and loading large volume bioprocess containers into Smartainer systems. Made of stainless still this is intended for use with the 2000L Smartainer II system.
Specifications
Capacity (Metric)
1500 L
For Use With (Equipment)
SV50160.04
Material
Stainless Steel (304)
Product Line
HyPerforma™, Smartainer™