HyPerforma™ Mixtainer™ Hardware, Conical Drum, 200L
HyPerforma™ Mixtainer™ Hardware, Conical Drum, 200L
Thermo Scientific™

HyPerforma™ Mixtainer™ Hardware, Conical Drum, 200L

Green features
The 200L Thermo Scientific™ Conical Drum top dispense, with clamps is utilized as part of the Thermo Scientific™ HyPerforma™ Mixtainer™자세히 알아보기
Have Questions?
카탈로그 번호수량
SV50517.13200 L
카탈로그 번호 SV50517.13
제품 가격(KRW)
-
견적 요청하기
수량:
200 L
The 200L Thermo Scientific™ Conical Drum top dispense, with clamps is utilized as part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications. Additional components of the system include the Thermo Scientific™ HyPerforma™ Mixtainer™ BioProcess Container (BPC) with an integrated magnetic stir bar, a magnetic stirring plate, a support plate and a dolly. The optimized support hardware allows mixing, moving, storage, shipping and discharge of the BPC contents while maintaining sterility of the contents. Standard products are available for sterile liquid-to-liquid or powder-to-liquid mixing.

Recommended for:
• Hydration of powdered media and buffers
• Sterile mixing of cell culture media with sera and reagents
• Sterile mixing of buffer solutions or other process fluids
• Pooling of sterile liquid fractions

사양
치수60 cm x 114 cm (23.5 in. x 44.75 in.)
그린 기능Fewer resources used and less waste, Fewer resources used and less waste
포트 세부 정보Bottom Port: 10.2 cm (4 in.)
제품라인HyPerforma, Mixtainer
수량200 L
용량(미터법)200 L
Unit Size200 L