Smart-Vue Pro Accessories
Smart-Vue Pro Accessories
Thermo Scientific™
Smart-Vue Pro Accessories
Customize your Smart-Vue Pro remote monitoring system with accessories to meet your specific needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
SVPADPPT1001Adapter2 or 3 Wire Generic PT100 Probes
SVPACCBAT002BatterySmart-Vue Pro Duo or Quatro Module
SVPADPPT1002AdapterSmart-Vue Classic PT100 Probes
Catalog number SVPADPPT1001
Price (USD)
-
Request A Quote
Type:
Adapter
For Use With (Equipment):
2 or 3 Wire Generic PT100 Probes
Choose from an assorment of Smart-Vue Pro accessories including power supplies, cables and mounting hardware, antenna and audible alarms to customize your remote monitoring system as well as ensure system integrity.
Compatible with Smart-Vue Pro remote monitoring system.
Specifications
CalibrationCalibration parameter are stored in the adapter. If calibrated sensors are connected a recalibration of the whole chain is recommended.
DescriptionSmart-Vue Pro adaptor for use with 2 or 3 wire generic PT100 probes. The adapter connects directly to a Smart-Vue Pro Duo or Quatro module
For Use With (Equipment)2 or 3 Wire Generic PT100 Probes
Length (English) Cable3.6 ft.
Measurement RangeDepends on the sensor connected at the adapter
Weight (Metric)150 g (Including shipping box)
TypeAdapter
Unit SizeEach