Thermo Scientific™
Smart-Vue Pro On-Premise Software and Subscription Services
Catalog number: SVPSWSM22000
Thermo Scientific™
Smart-Vue Pro On-Premise Software and Subscription Services
Catalog number: SVPSWSM22000
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions the system will automatically notify the user based on prescribed notification preferences and allows you to document notes around each specific event for audit purposes. The floorplan view and local audio visual alarms will make it easy to locate alarming equipment during the critical recovery times. The on-premise solution maintains your Smart-Vue Pro data locally on your site's server.
Catalog Number
SVPSWSM22000
Unit Size
Each
Description
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 2000 PTS
Price (USD)
Full specifications
DescriptionSmart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 2000 PTS
For Use With (Equipment)Smart-Vue Pro System
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Pro Web application on-premise license up to 5 measurement points
Price (USD)
Full specifications
DescriptionSmart-Vue Pro Web application on-premise license up to 5 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Pro Web application on-premise license up to 20 measurement points
Price (USD)
Full specifications
DescriptionSmart-Vue Pro Web application on-premise license up to 20 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Pro Web application on-premise license up to 50 measurement points
Price (USD)
Full specifications
DescriptionSmart-Vue Pro Web application on-premise license up to 50 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Pro Web application on-premise license up to 100 measurement points
Price (USD)
Full specifications
DescriptionSmart-Vue Pro Web application on-premise license up to 100 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Pro Web application on-premise license up to 250 measurement points
Price (USD)
Full specifications
DescriptionSmart-Vue Pro Web application on-premise license up to 250 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Pro Web application on-premise license up to 500 measurement points
Price (USD)
Full specifications
DescriptionSmart-Vue Pro Web application on-premise license up to 500 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Pro Web application on-premise license up to 1000 measurement points
Price (USD)
Full specifications
DescriptionSmart-Vue Pro Web application on-premise license up to 1000 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Pro Web application on-premise license up to 1500 measurement points
Price (USD)
Full specifications
DescriptionSmart-Vue Pro Web application on-premise license up to 1500 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Smart-Vue Pro Web application on-premise license up to 2000 measurement points
Price (USD)
Full specifications
DescriptionSmart-Vue Pro Web application on-premise license up to 2000 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Showing 10 of 26Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
SVPSWSM22000Full specifications
EachSmart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 2000 PTSRequest A Quote
DescriptionSmart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 2000 PTS
For Use With (Equipment)Smart-Vue Pro System
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
SVPSWPS20005Full specifications
EachSmart-Vue Pro Web application on-premise license up to 5 measurement pointsRequest A Quote
DescriptionSmart-Vue Pro Web application on-premise license up to 5 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
SVPSWPS20020Full specifications
EachSmart-Vue Pro Web application on-premise license up to 20 measurement pointsRequest A Quote
DescriptionSmart-Vue Pro Web application on-premise license up to 20 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
SVPSWPS20050Full specifications
EachSmart-Vue Pro Web application on-premise license up to 50 measurement pointsRequest A Quote
DescriptionSmart-Vue Pro Web application on-premise license up to 50 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
SVPSWPS20100Full specifications
EachSmart-Vue Pro Web application on-premise license up to 100 measurement pointsRequest A Quote
DescriptionSmart-Vue Pro Web application on-premise license up to 100 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
SVPSWPS20250Full specifications
EachSmart-Vue Pro Web application on-premise license up to 250 measurement pointsRequest A Quote
DescriptionSmart-Vue Pro Web application on-premise license up to 250 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
SVPSWPS20500Full specifications
EachSmart-Vue Pro Web application on-premise license up to 500 measurement pointsRequest A Quote
DescriptionSmart-Vue Pro Web application on-premise license up to 500 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
SVPSWPS21000Full specifications
EachSmart-Vue Pro Web application on-premise license up to 1000 measurement pointsRequest A Quote
DescriptionSmart-Vue Pro Web application on-premise license up to 1000 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
SVPSWPS21500Full specifications
EachSmart-Vue Pro Web application on-premise license up to 1500 measurement pointsRequest A Quote
DescriptionSmart-Vue Pro Web application on-premise license up to 1500 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
SVPSWPS22000Full specifications
EachSmart-Vue Pro Web application on-premise license up to 2000 measurement pointsRequest A Quote
DescriptionSmart-Vue Pro Web application on-premise license up to 2000 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Unit SizeEach
Showing 10 of 26Show more
  • Intuitive dashboard simplifies daily usage.
  • Optional floor plan and sites views.
  • Data can be exported with a click for reporting and customizable graphs.
  • Web application on Cloud or customer server.
  • Instant access to alarms, readings, and graphs.
  • Remote data access.
  • Flexible organization by site, department user, equipment, and criticality.
  • Audit trail and secure-level access control to support 21CFR Part 11
Figures