Sensititre™ SWIN™ Software System
Thermo Scientific™

Sensititre™ SWIN™ Software System

Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Thermo Scientific™ Sensititre™ SWIN™ Software System.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripción
SW1302Monitor de pantalla táctil de 22 in
SW1202Software del módulo de epidemiología
SW4000Software completo
SW1100Etiquetas, código de barras
SW1301Monitor de pantalla táctil de 17 in
SW516Interfaz de Bruker
Número de catálogo SW1302
Precio (MXN)
-
Descripción:
Monitor de pantalla táctil de 22 in
Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Thermo Scientific™ Sensititre™ SWIN™ Software System. Simple touch screen navigation puts results at your fingertips while drop-down fields eliminate manual entry errors for all manual, semi-automated and fully automated reads.
Consolidate your entire test program on a single software platform while enhancing data entry and reporting with the Sensititre SWIN Software System.
  • Customizable Expert System provides three-tiered expert messages: Information, Modification (based on FDA, CLSI and EUCAST recommendations) and Warning Levels
  • Quality control module easily manages MIC results and broth information for inspection purposes
  • Automated alerts and report functionality enhance workflow and expedite release of results

Includes:
Microsoft Windows PC including dual hard drives, internal DVD burner, internal modem, barcode scanner, laser printer, remote access software, touchscreen monitor and SWIN Software.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificaciones
DescripciónMonitor de pantalla táctil de 22 in
Tipo de productoMicrobiology Media
Unit SizeEach