WebREMS™ Web-based Real-time Environmental Monitoring System
WebREMS™ Web-based Real-time Environmental Monitoring System
Actual product may vary
Thermo Scientific™
WebREMS™ Web-based Real-time Environmental Monitoring System
Be sure personnel exposed to ionizing radiation don't exceed allowable dose limits and receive appropriate decontamination treatment with a web-based EPD monitoring system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Application)
SYS170860008
also known as SYS/1/70860/008
Additional Site LicencesAdditional Sites
SYS170860002
also known as SYS/1/70860/002
Single Site License0 to 50 Devices
SYS170860003
also known as SYS/1/70860/003
Single Site License0 to 100 Devices
SYS170860004
also known as SYS/1/70860/004
Single Site License0 to 200 Devices
SYS170860005
also known as SYS/1/70860/005
Single Site License0 to 750 Devices
SYS170860006
also known as SYS/1/70860/006
Single Site License0 to 1500 Devices
SYS170860011
also known as SYS/1/70860/011
Dosimeter Device LicenseUnlimited
SYS170860012
also known as SYS/1/70860/012
Site LicenseUnlimited
Catalog number SYS170860008
also known as SYS/1/70860/008
Price (MXN)
-
Product Type:
Additional Site Licences
For Use With (Application):
Additional Sites
Personnel working in environments where they are exposed to ionizing radiation must have their exposure accurately monitored, recorded and reported. The Thermo Scientific™ webREMS™ Web-based Real-time Environmental Monitoring System tracks vital data from your electronic personal dosimeter (EPD) population including worker dosimeter visits, training status, dosimeters status, tasks\permits and radiation dose summary. The system also performs the EPD issue and return process. With this easy-to-use web-based graphical interface, accurate data and full reporting capabilities are at your fingertips.
Thermo Scientific webREMS Web-based Real-Time Environmental Monitoring System for monitoring and reporting personnel radiation exposure has built-in reporting capability and allows system administration from anywhere in the world.
Features
  • Client/server web-based application designed to track and monitor personnel radiation exposure.
  • Facilitates distributed access control through simple to manage electronic personal dosimeter (EPD) issue and return stations through an easy to use web browser, utilizing Microsoft™ Internet Explorer™ (IE).
  • Facilitates data management of personnel radiological dose limits, training requirements and authorization against Tasks.
  • Integrates with Thermo Scientific™ ViewPoint™ Enterprise to provide real-time monitoring of personnel exposed to ionizing radiation.
  • Integrates with Thermo Scientific™ webALARA web-based As Low As Reasonably Achievable permit management system.
  • Provides reporting capabilities of all visits performed using the system, recording and reporting the dose recorded against personnel and tasks/permits over one or more configurable time periods.
  • Facilitates simple, quick integration with existing data through a built-in import process for personnel, dosimeter calibration expiry and training data.
  • Scalable such that webREMS may be installed and operated from a single laptop PC up to an enterprise server installation providing access anywhere in the world.
  • Secure (HTTPS) access over public internet with password protected system access via ViewPoint Authentication Service.
  • Typical uses include nuclear power plants, government labs, federal agencies, security and military.
Requires
Microsoft™ Windows™ PC with Internet Explorer™ (IE) and Internet Information Services (IIS).
Thermo Scientific™ Electronic Personal Dosimeter (EPD), Beta/Gamma, Gamma, Neutron or Fast-Neutron.
Thermo Scientific™ IRDA Reader to use Access Control.
Specifications
CompatibilityThermo Scientific EPD Mk2, EPD-N, EPD-N2
DescriptionAdditional site licenses
For Use With (Application)Additional Sites
Operating SystemMicrosoft Windows 2008, 2008 R2, XP, 7 (32 and 64 bit)
Product TypeAdditional Site Licences
Unit SizeEach