Specifications
Boiling Point111°C
CAS108-88-3
Chemical Name or MaterialToluene
IUPAC Nametoluene
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
MDL NumberMFCD00008512
Melting Point-95°C
Molecular FormulaC7H8
PackagingMetal Drum
SMILESCC1=CC=CC=C1
Molecular Weight (g/mol)92.14
Quantity200 L
Unit SizeEach