This product ships via ground from US northeast.
Thermo Scientific™

Nitric Acid, ICP-OES, for Trace Metal Analysis, Thermo Scientific™

Catalog number: T003092500CS
Thermo Scientific™

Nitric Acid, ICP-OES, for Trace Metal Analysis, Thermo Scientific™

Catalog number: T003092500CS
 
Catalog Number
T003092500CS
also known as T003092500
Unit Size
Case of,4
Quantity
2.5L
Price (USD)
356.10
Your Price:
Availability
***
Full specifications
Quantity
Aluminum (Al)≤ 30 ppb
Chemical Name or MaterialNitric Acid
Color10 max. (APHA)
Quantity2.5L
CAS7697-37-2
AppearanceClear, colorless liquid
Arsenic (As)≤ 5 ppb
Barium (Ba)≤ 1 ppb
Bismuth (Bi)≤ 1 ppb
Boron (B)≤ 4 ppb
Purity Grade NotesFor trace metal analysis
Cadmium (Cd)≤ 1 ppb
Molecular FormulaHNO3
Calcium (Ca)≤ 50 ppb
Cobalt (Co)≤ 1 ppb
Chromium (Cr)≤ 10 ppb
Chloride≤ 0.05 ppm
Synonymhydrogen nitrate
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES[N+](=O)(O)[O-]
IUPAC Namenitric acid
Density1.42
Molecular Weight (g/mol)63.01
PubChem CID944
ChEBICHEBI:48107
Copper (Cu)≤ 1 ppb
Percent Purity69.0 - 70.0%
GradeICP-OES
Gold (Au)≤ 4 ppb
Heavy Metals (as Pb)100 ppb max.
Iron (Fe)≤ 10 ppb
Lead (Pb)≤ 0.5 ppb
Lithium (Li)≤ 1 ppb
Manganese (Mn)≤ 1 ppb
Magnesium (Mg)≤ 7 ppb
Mercury (Hg)≤ 0.5 ppb
Molybdenum (Mo)≤ 5 ppb
Nickel (Ni)≤ 1 ppb
PackagingPoly coated glass bottle
Phosphate≤ 0.10 ppm
Potassium (K)≤ 5 ppb
Residue after Ignition≤ 2 ppm
Silver (Ag)≤ 1 ppb
Sodium (Na)≤ 200 ppb
Strontium (Sr)≤ 1 ppb
Sulfate≤ 0.4 ppm
Tin (Sn)≤ 5 ppb
Vanadium (V)≤ 1 ppb
Zinc (Zn)≤ 5 ppb
Zirconium (Zr)≤ 1 ppb
Unit SizeCase of,4
Catalog Number
Unit Size
Each
Quantity
500mL
Price (USD)
79.00
Your Price:
Availability
***
Full specifications
Quantity
Aluminum (Al)≤ 30 ppb
Chemical Name or MaterialNitric Acid
Color10 max. (APHA)
Quantity500mL
CAS7697-37-2
AppearanceClear, colorless liquid
Arsenic (As)≤ 5 ppb
Barium (Ba)≤ 1 ppb
Bismuth (Bi)≤ 1 ppb
Boron (B)≤ 4 ppb
Purity Grade NotesFor trace metal analysis
Cadmium (Cd)≤ 1 ppb
Molecular FormulaHNO3
Calcium (Ca)≤ 50 ppb
Cobalt (Co)≤ 1 ppb
Chromium (Cr)≤ 10 ppb
Chloride≤ 0.05 ppm
Synonymhydrogen nitrate
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES[N+](=O)(O)[O-]
IUPAC Namenitric acid
Density1.42
Molecular Weight (g/mol)63.01
PubChem CID944
ChEBICHEBI:48107
Copper (Cu)≤ 1 ppb
Percent Purity69.0 - 70.0%
GradeICP-OES
Gold (Au)≤ 4 ppb
Heavy Metals (as Pb)100 ppb max.
Iron (Fe)≤ 10 ppb
Lead (Pb)≤ 0.5 ppb
Lithium (Li)≤ 1 ppb
Manganese (Mn)≤ 1 ppb
Magnesium (Mg)≤ 7 ppb
Mercury (Hg)≤ 0.5 ppb
Molybdenum (Mo)≤ 5 ppb
Nickel (Ni)≤ 1 ppb
PackagingPoly coated glass bottle
Phosphate≤ 0.10 ppm
Potassium (K)≤ 5 ppb
Residue after Ignition≤ 2 ppm
Silver (Ag)≤ 1 ppb
Sodium (Na)≤ 200 ppb
Strontium (Sr)≤ 1 ppb
Sulfate≤ 0.4 ppm
Tin (Sn)≤ 5 ppb
Vanadium (V)≤ 1 ppb
Zinc (Zn)≤ 5 ppb
Zirconium (Zr)≤ 1 ppb
Unit SizeEach
Catalog Number
T003090500CS
also known as T003090500
Unit Size
Case of,6
Quantity
500mL
Price (USD)
426.60
Your Price:
Availability
***
Full specifications
Quantity
Aluminum (Al)≤ 30 ppb
Chemical Name or MaterialNitric Acid
Color10 max. (APHA)
Quantity500mL
CAS7697-37-2
AppearanceClear, colorless liquid
Arsenic (As)≤ 5 ppb
Barium (Ba)≤ 1 ppb
Bismuth (Bi)≤ 1 ppb
Boron (B)≤ 4 ppb
Purity Grade NotesFor trace metal analysis
Cadmium (Cd)≤ 1 ppb
Molecular FormulaHNO3
Calcium (Ca)≤ 50 ppb
Cobalt (Co)≤ 1 ppb
Chromium (Cr)≤ 10 ppb
Chloride≤ 0.05 ppm
Synonymhydrogen nitrate
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES[N+](=O)(O)[O-]
IUPAC Namenitric acid
Density1.42
Molecular Weight (g/mol)63.01
PubChem CID944
ChEBICHEBI:48107
Copper (Cu)≤ 1 ppb
Percent Purity69.0 - 70.0%
GradeICP-OES
Gold (Au)≤ 4 ppb
Heavy Metals (as Pb)100 ppb max.
Iron (Fe)≤ 10 ppb
Lead (Pb)≤ 0.5 ppb
Lithium (Li)≤ 1 ppb
Manganese (Mn)≤ 1 ppb
Magnesium (Mg)≤ 7 ppb
Mercury (Hg)≤ 0.5 ppb
Molybdenum (Mo)≤ 5 ppb
Nickel (Ni)≤ 1 ppb
PackagingPoly coated glass bottle
Phosphate≤ 0.10 ppm
Potassium (K)≤ 5 ppb
Residue after Ignition≤ 2 ppm
Silver (Ag)≤ 1 ppb
Sodium (Na)≤ 200 ppb
Strontium (Sr)≤ 1 ppb
Sulfate≤ 0.4 ppm
Tin (Sn)≤ 5 ppb
Vanadium (V)≤ 1 ppb
Zinc (Zn)≤ 5 ppb
Zirconium (Zr)≤ 1 ppb
Unit SizeCase of,6
Catalog Number
Unit Size
Each
Quantity
2.5L
Price (USD)
99.00
Your Price:
Availability
***
Full specifications
Quantity
Aluminum (Al)≤ 30 ppb
Chemical Name or MaterialNitric Acid
Color10 max. (APHA)
Quantity2.5L
CAS7697-37-2
AppearanceClear, colorless liquid
Arsenic (As)≤ 5 ppb
Barium (Ba)≤ 1 ppb
Bismuth (Bi)≤ 1 ppb
Boron (B)≤ 4 ppb
Purity Grade NotesFor trace metal analysis
Cadmium (Cd)≤ 1 ppb
Molecular FormulaHNO3
Calcium (Ca)≤ 50 ppb
Cobalt (Co)≤ 1 ppb
Chromium (Cr)≤ 10 ppb
Chloride≤ 0.05 ppm
Synonymhydrogen nitrate
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES[N+](=O)(O)[O-]
IUPAC Namenitric acid
Density1.42
Molecular Weight (g/mol)63.01
PubChem CID944
ChEBICHEBI:48107
Copper (Cu)≤ 1 ppb
Percent Purity69.0 - 70.0%
GradeICP-OES
Gold (Au)≤ 4 ppb
Heavy Metals (as Pb)100 ppb max.
Iron (Fe)≤ 10 ppb
Lead (Pb)≤ 0.5 ppb
Lithium (Li)≤ 1 ppb
Manganese (Mn)≤ 1 ppb
Magnesium (Mg)≤ 7 ppb
Mercury (Hg)≤ 0.5 ppb
Molybdenum (Mo)≤ 5 ppb
Nickel (Ni)≤ 1 ppb
PackagingPoly coated glass bottle
Phosphate≤ 0.10 ppm
Potassium (K)≤ 5 ppb
Residue after Ignition≤ 2 ppm
Silver (Ag)≤ 1 ppb
Sodium (Na)≤ 200 ppb
Strontium (Sr)≤ 1 ppb
Sulfate≤ 0.4 ppm
Tin (Sn)≤ 5 ppb
Vanadium (V)≤ 1 ppb
Zinc (Zn)≤ 5 ppb
Zirconium (Zr)≤ 1 ppb
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)AvailabilityQuantity
T003092500CS
also known as T003092500
Full specifications
Case of,42.5L
356.10
Your Price:
***
Aluminum (Al)≤ 30 ppb
Chemical Name or MaterialNitric Acid
Color10 max. (APHA)
Quantity2.5L
CAS7697-37-2
AppearanceClear, colorless liquid
Arsenic (As)≤ 5 ppb
Barium (Ba)≤ 1 ppb
Bismuth (Bi)≤ 1 ppb
Boron (B)≤ 4 ppb
Purity Grade NotesFor trace metal analysis
Cadmium (Cd)≤ 1 ppb
Molecular FormulaHNO3
Calcium (Ca)≤ 50 ppb
Cobalt (Co)≤ 1 ppb
Chromium (Cr)≤ 10 ppb
Chloride≤ 0.05 ppm
Synonymhydrogen nitrate
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES[N+](=O)(O)[O-]
IUPAC Namenitric acid
Density1.42
Molecular Weight (g/mol)63.01
PubChem CID944
ChEBICHEBI:48107
Copper (Cu)≤ 1 ppb
Percent Purity69.0 - 70.0%
GradeICP-OES
Gold (Au)≤ 4 ppb
Heavy Metals (as Pb)100 ppb max.
Iron (Fe)≤ 10 ppb
Lead (Pb)≤ 0.5 ppb
Lithium (Li)≤ 1 ppb
Manganese (Mn)≤ 1 ppb
Magnesium (Mg)≤ 7 ppb
Mercury (Hg)≤ 0.5 ppb
Molybdenum (Mo)≤ 5 ppb
Nickel (Ni)≤ 1 ppb
PackagingPoly coated glass bottle
Phosphate≤ 0.10 ppm
Potassium (K)≤ 5 ppb
Residue after Ignition≤ 2 ppm
Silver (Ag)≤ 1 ppb
Sodium (Na)≤ 200 ppb
Strontium (Sr)≤ 1 ppb
Sulfate≤ 0.4 ppm
Tin (Sn)≤ 5 ppb
Vanadium (V)≤ 1 ppb
Zinc (Zn)≤ 5 ppb
Zirconium (Zr)≤ 1 ppb
Unit SizeCase of,4
T003090500Full specifications
Each500mL
79.00
Your Price:
***
Aluminum (Al)≤ 30 ppb
Chemical Name or MaterialNitric Acid
Color10 max. (APHA)
Quantity500mL
CAS7697-37-2
AppearanceClear, colorless liquid
Arsenic (As)≤ 5 ppb
Barium (Ba)≤ 1 ppb
Bismuth (Bi)≤ 1 ppb
Boron (B)≤ 4 ppb
Purity Grade NotesFor trace metal analysis
Cadmium (Cd)≤ 1 ppb
Molecular FormulaHNO3
Calcium (Ca)≤ 50 ppb
Cobalt (Co)≤ 1 ppb
Chromium (Cr)≤ 10 ppb
Chloride≤ 0.05 ppm
Synonymhydrogen nitrate
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES[N+](=O)(O)[O-]
IUPAC Namenitric acid
Density1.42
Molecular Weight (g/mol)63.01
PubChem CID944
ChEBICHEBI:48107
Copper (Cu)≤ 1 ppb
Percent Purity69.0 - 70.0%
GradeICP-OES
Gold (Au)≤ 4 ppb
Heavy Metals (as Pb)100 ppb max.
Iron (Fe)≤ 10 ppb
Lead (Pb)≤ 0.5 ppb
Lithium (Li)≤ 1 ppb
Manganese (Mn)≤ 1 ppb
Magnesium (Mg)≤ 7 ppb
Mercury (Hg)≤ 0.5 ppb
Molybdenum (Mo)≤ 5 ppb
Nickel (Ni)≤ 1 ppb
PackagingPoly coated glass bottle
Phosphate≤ 0.10 ppm
Potassium (K)≤ 5 ppb
Residue after Ignition≤ 2 ppm
Silver (Ag)≤ 1 ppb
Sodium (Na)≤ 200 ppb
Strontium (Sr)≤ 1 ppb
Sulfate≤ 0.4 ppm
Tin (Sn)≤ 5 ppb
Vanadium (V)≤ 1 ppb
Zinc (Zn)≤ 5 ppb
Zirconium (Zr)≤ 1 ppb
Unit SizeEach
T003090500CS
also known as T003090500
Full specifications
Case of,6500mL
426.60
Your Price:
***
Aluminum (Al)≤ 30 ppb
Chemical Name or MaterialNitric Acid
Color10 max. (APHA)
Quantity500mL
CAS7697-37-2
AppearanceClear, colorless liquid
Arsenic (As)≤ 5 ppb
Barium (Ba)≤ 1 ppb
Bismuth (Bi)≤ 1 ppb
Boron (B)≤ 4 ppb
Purity Grade NotesFor trace metal analysis
Cadmium (Cd)≤ 1 ppb
Molecular FormulaHNO3
Calcium (Ca)≤ 50 ppb
Cobalt (Co)≤ 1 ppb
Chromium (Cr)≤ 10 ppb
Chloride≤ 0.05 ppm
Synonymhydrogen nitrate
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES[N+](=O)(O)[O-]
IUPAC Namenitric acid
Density1.42
Molecular Weight (g/mol)63.01
PubChem CID944
ChEBICHEBI:48107
Copper (Cu)≤ 1 ppb
Percent Purity69.0 - 70.0%
GradeICP-OES
Gold (Au)≤ 4 ppb
Heavy Metals (as Pb)100 ppb max.
Iron (Fe)≤ 10 ppb
Lead (Pb)≤ 0.5 ppb
Lithium (Li)≤ 1 ppb
Manganese (Mn)≤ 1 ppb
Magnesium (Mg)≤ 7 ppb
Mercury (Hg)≤ 0.5 ppb
Molybdenum (Mo)≤ 5 ppb
Nickel (Ni)≤ 1 ppb
PackagingPoly coated glass bottle
Phosphate≤ 0.10 ppm
Potassium (K)≤ 5 ppb
Residue after Ignition≤ 2 ppm
Silver (Ag)≤ 1 ppb
Sodium (Na)≤ 200 ppb
Strontium (Sr)≤ 1 ppb
Sulfate≤ 0.4 ppm
Tin (Sn)≤ 5 ppb
Vanadium (V)≤ 1 ppb
Zinc (Zn)≤ 5 ppb
Zirconium (Zr)≤ 1 ppb
Unit SizeCase of,6
T003092500Full specifications
Each2.5L
99.00
Your Price:
***
Aluminum (Al)≤ 30 ppb
Chemical Name or MaterialNitric Acid
Color10 max. (APHA)
Quantity2.5L
CAS7697-37-2
AppearanceClear, colorless liquid
Arsenic (As)≤ 5 ppb
Barium (Ba)≤ 1 ppb
Bismuth (Bi)≤ 1 ppb
Boron (B)≤ 4 ppb
Purity Grade NotesFor trace metal analysis
Cadmium (Cd)≤ 1 ppb
Molecular FormulaHNO3
Calcium (Ca)≤ 50 ppb
Cobalt (Co)≤ 1 ppb
Chromium (Cr)≤ 10 ppb
Chloride≤ 0.05 ppm
Synonymhydrogen nitrate
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES[N+](=O)(O)[O-]
IUPAC Namenitric acid
Density1.42
Molecular Weight (g/mol)63.01
PubChem CID944
ChEBICHEBI:48107
Copper (Cu)≤ 1 ppb
Percent Purity69.0 - 70.0%
GradeICP-OES
Gold (Au)≤ 4 ppb
Heavy Metals (as Pb)100 ppb max.
Iron (Fe)≤ 10 ppb
Lead (Pb)≤ 0.5 ppb
Lithium (Li)≤ 1 ppb
Manganese (Mn)≤ 1 ppb
Magnesium (Mg)≤ 7 ppb
Mercury (Hg)≤ 0.5 ppb
Molybdenum (Mo)≤ 5 ppb
Nickel (Ni)≤ 1 ppb
PackagingPoly coated glass bottle
Phosphate≤ 0.10 ppm
Potassium (K)≤ 5 ppb
Residue after Ignition≤ 2 ppm
Silver (Ag)≤ 1 ppb
Sodium (Na)≤ 200 ppb
Strontium (Sr)≤ 1 ppb
Sulfate≤ 0.4 ppm
Tin (Sn)≤ 5 ppb
Vanadium (V)≤ 1 ppb
Zinc (Zn)≤ 5 ppb
Zirconium (Zr)≤ 1 ppb
Unit SizeEach
Showing 4 of 4
  • Suitable for trace metal analysis by ICP-OES
  • Tested for 34 metal impurities at ppb levels
  • Also meets general ACS testing specifications
  • Ideal for analysis of environmental samples
  • Offered in 500mL and 2.5L poly coated glass bottles
RUO – Research Use Only

Figures

Frequently asked questions (FAQs)

Citations & References